Skip to Content
Merck

Skip To

A7638

Bovine Serum Albumin

lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥99% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
blocking: suitable, microbiological culture: suitable
Impurities:
essentially globulin free
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥99% (agarose gel electrophoresis)

form

lyophilized powder

purified by

cold ethanol and heatshock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

blocking: suitable, microbiological culture: suitable

impurities

essentially globulin free

pH

7

solubility

water: soluble (50 mg/ml)

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Purified by a combination of heat shock and ethanol fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3983A0281A3608
technique(s)

blocking: suitable, microbiological culture: suitable

technique(s)

western blot: suitable

technique(s)

enzyme immunoassay: suitable

technique(s)

immunoprecipitation (IP): suitable

origin

USA origin

origin

New Zealand origin

origin

USA origin

origin

New Zealand origin

assay

≥99% (agarose gel electrophoresis)

assay

≥98% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A7638-100G04061833385586
A7638-25G04061833385838
A7638-250MG04061833385821
A7638-10G04061833385661
A7638-5G04061833385869
A7638-1G04061833385777

Questions

Reviews

No rating value

Active Filters