Skip to Content
Merck

M1636

Myosin, Calcium activated from rabbit muscle

buffered aqueous glycerol solution, 0.5-1.5 units/mg protein (biuret)

Synonym(s):

Myosin adenosinetriphosphatase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Form:
buffered aqueous glycerol solution
Biological source:
rabbit muscle
Mol wt:
heavy chain ~200 kDa (each), light chain 15-20 kDa (each), ~480 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit muscle

Quality Level

form

buffered aqueous glycerol solution

specific activity

0.5-1.5 units/mg protein (biuret)

mol wt

heavy chain ~200 kDa (each), light chain 15-20 kDa (each), ~480 kDa

technique(s)

ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Interacts with actin in muscle and non-muscle cells; involved in neurosensory function, vesicle trafficking, determinant partitioning, and cortical function.
Myosin interacts with actin in muscle and non-muscle cells. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Myosin molecules spontaneously assemble into filaments in solutions of physiologic ionic strength and pH. Thick filament consists mainly of myosin molecules. Myosin is activated by the enzyme ATPase. This activation is the immediate source of the free energy that drives muscle contraction. It binds to the polymerized form of actin, the major constituent of the thin filament. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. These are designated as type I (slow-twitch) and type II (fast-twitch). Myosin is one of three classes of cytoskeletal motor proteins that have been identified. The others are kinesins and dyneins. These three types of proteins are thought to be responsible for the many movements that occur in cells. Myosin has been shown to be involved in neurosensory function, vesicle trafficking, determinant partitioning, and cortical function.
Myosin interacts with actin in muscle and non-muscle cells. Myosin is one of three classes of cytoskeletal motor proteins that have been identified. The others are kinesins and dyneins. These three types of proteins are thought to be responsible for the many movements that occur in cells. Myosin has been shown to be involved in neurosensory function, vesicle trafficking, determinant partitioning, and cortical function.

Physical form

Solution in 50% glycerol containing 0.6 M potassium chloride and 10 mM potassium phosphate buffer, pH 6.8. Note: Solution may be hazy.

Other Notes

One unit will liberate 1.0 μmole of inorganic phosphorus from ATP per min at pH 9.0 at 25 °C in the presence of calcium.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lichuang Cao et al.
Food chemistry, 356, 129655-129655 (2021-04-09)
Phosphorylation of myosin regulatory light chain (MRLC) can regulate muscle contraction and thus affect actomyosin dissociation and meat quality. The objective of this study was to explore the mechanism by how MRLC phosphorylation regulates actomyosin dissociation and thus develop strategies
Brandon M Hoover et al.
Journal of pharmaceutical sciences, 109(1), 452-463 (2019-10-12)
Nanoparticle tracking analysis (NTA) is an important technique for measuring hydrodynamic size of globular biological particles including liposomes and viruses. Less attention has been paid to NTA of rod-like particles, despite their considerable interest. For example, amyloid fibrils and protofibrils
E Bischoff et al.
Molecular microbiology, 35(5), 1005-1016 (2000-03-11)
Four large multigene families have been described in Plasmodium falciparum malaria parasites (var, rif, stevor and Pf60). var and rif genes code for erythrocyte surface proteins and undergo clonal antigenic variation. We report here the characterization of the first Pf60



Global Trade Item Number

SKUGTIN
M1636-5MG04061833272794
M1636-10MG04061833272770
M1636-50MG04061833272787