Skip to Content
Merck
All Photos(6)

Documents

105228

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol

≥99%

Synonym(s):

HFP, Hexafluoroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CHOH
CAS Number:
Molecular Weight:
168.04
Beilstein:
1841007
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

refractive index

n20/D 1.275 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.596 g/mL at 25 °C (lit.)

SMILES string

OC(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H

InChI key

BYEAHWXPCBROCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,1,3,3,3-Hexafluoro-2-propanol is a solution-phase peptide chemistry solvent. This fluorinated polar solvent of high ionizing power facilitates Friedel–Crafts-type reactions, using covalent reagents in the absence of a Lewis acid catalyst. It also enhances the efficiency of rhodium(I)-catalyzed [4+2] intramolecular cycloaddition of ether-tethered alkynyl dienes and [5+2] cycloaddition of alkynyl vinylcyclopropanes. 1,1,1,3,3,3-Hexafluoro-2-propanol clusters catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide.

Application

1,1,1,3,3,3-Hexafluoro-2-propanol was used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Central nervous system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rita Sobreiro-Almeida et al.
Materials science & engineering. C, Materials for biological applications, 103, 109866-109866 (2019-07-28)
Kidney diseases are recognized as a major health problem, which affect 10% of the population. Because currently available therapies have many limitations, some tissue engineering strategies have been emerging as promising approaches in this field. In this work, porcine kidneys
Hexafluoroalcohol-functionalized Methacrylate Monomers for Lithographic/Nanopatterning Materials.
Sanders DP, et al.
Material Matters, 6(1), 1-7 (2011)
Akio Saito et al.
The Journal of organic chemistry, 71(17), 6437-6443 (2006-08-12)
In fluorinated alcohols, the cationic Rh(I) species, which is derived from [Rh(COD)Cl]2 and AgSbF6, efficiently catalyzed intramolecular [4+2] cycloaddition reactions of ester-tethered 1,3-diene-8-yne derivatives. The catalytic system was also effective in intramolecular [5+2] cycloaddition reactions of ester-tethered omega-alkynyl vinylcyclopropane compounds.
Kinetic Studies of Olefin Epoxidation with Hydrogen Peroxide in 1, 1, 1, 3, 3, 3-Hexafluoro-2-propanol Reveal a Crucial Catalytic Role for Solvent Clusters.
Berkessel A and Adrio JA.
Advanced Synthesis & Catalysis, 346(2-3), 275-280 (2004)
K Halverson et al.
Biochemistry, 29(11), 2639-2644 (1990-03-20)
The amyloid beta-protein (1-42) is a major constituent of the abnormal extracellular amyloid plaque that characterizes the brains of victims of Alzheimer's disease. Two peptides, with sequences derived from the previously unexplored C-terminal region of the beta-protein, beta 26-33 (H2N-SNKGAIIG-CO2H)

Articles

To keep pace with Moore′s Law, there is a continuing need in the semiconductor industry to achieve higher circuit density in microelectronic devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service