Skip to Content
Merck
All Photos(1)

Key Documents

90362

Sigma-Aldrich

Triethylammonium phosphate solution

BioXtra, 1 M in H2O

Synonym(s):

Buffer solution 1 M pH 3.0, TEAP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

form

liquid

concentration

1 M in H2O

refractive index

n20/D 1.360

pH

2.9-3.1 (25 °C)

density

1.05 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

application(s)

diagnostic assay manufacturing

SMILES string

OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CC

InChI

1S/C6H15N.H3O4P/c1-4-7(5-2)6-3;1-5(2,3)4/h4-6H2,1-3H3;(H3,1,2,3,4)

InChI key

UNXNGGMLCSMSLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Triethylammonium phosphate (TEAP) buffers are highly efficient solvents, that can be used for reversed-phase high-performance liquid chromatography (RP-HPLC).

Other Notes

Mobile phase (15 mM) for reversed phase HPLC of amino acids, peptides and proteins

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.T.W. Hearn et al.
Journal of Chromatography A, 218, 497-497 (1981)
J.E. Rivier
Journal of Liquid Chromatography, 1, 343-343 (1979)
Evaluation of triethylammonium phosphate and formate-acetonitrile mixtures as eluents for high-performance gel permeation chromatography
Rivier JE
Journal of Chromatography A, 202(2), 211-222 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service