Skip to Content
Merck
All Photos(2)

Key Documents

23186

Supelco

Fmoc chloride

≥99.0%, for HPLC derivatization, LiChropur

Synonym(s):

9-Fluorenylmethoxycarbonyl chloride, 9-Fluorenylmethyl chloroformate, Fmoc-Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO2
CAS Number:
Molecular Weight:
258.70
Beilstein:
2279177
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

Product Name

Fmoc chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

mp

61-64 °C
62-64 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C15H11ClO2/c16-15(17)18-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9H2

InChI key

IRXSLJNXXZKURP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc chloride is a derivatizing agent.

Application

Fmoc chloride may be used as derivatizing reagent in pre-column derivatization of the biogenic amines, resolved using HPLC technique.
Reagent for derivatizing amino acids for HPLC amino acid analysis and for preparing N-Fmoc amino acids for solid-phase peptide synthesis.

Other Notes

Reagent for precolumn derivatization of amines for HPLC and fluorescent detection (abs.: 265 nm, em.: 315 nm); Used for capillary electrophoresis

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.V. Sancho et al.
Journal of Chromatography A, 678, 59-59 (1994)
K C Chan et al.
Journal of chromatography. A, 653(1), 93-97 (1993-10-29)
Laser-induced fluorescence (LIF) was applied to the detection of 9-fluorenylmethyl chloroformate (FMOC-Cl) derivatized amino acids separated by capillary electrophoresis. Fluorescence excitation was provided by a pulsed, KrF laser operating at 248 nm. A limit of detection of 5 x 10(-10)
J. Kirschbaum et al.
Journal of Chromatography A, 661, 193-193 (1994)
S. Einarsson et al.
Analytical Chemistry, 58, 1638-1638 (1986)
Guoying Cao et al.
The Journal of antimicrobial chemotherapy, 62(5), 1009-1014 (2008-09-04)
The purpose of this study was to develop a specific, sensitive, accurate and reproducible high-performance liquid chromatographic (HPLC) method to measure polymyxin B in human plasma. Derivatization of polymyxin B with fluorescent 9-fluorenylmethyl chloroformate (FMOC-Cl) was performed in the same

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service