Skip to Content
Merck
All Photos(1)

Key Documents

31457

Sigma-Aldrich

Sodium tetraborate decahydrate

puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Agency

reag. ISO

Assay

≥99.5%

form

powder or crystals

pH

9.0-9.6 (20 °C, 5%)

density

1.73 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Pb: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium tetraborate decahydrate may be used in the following studies:
  • Modified carbazole method for the quantitive determination of hexuronic acids.
  • Quantitative estimation of aminoglycoside antibiotics (dihydrostreptomycin and streptomycin) by capillary electrophoresis coupled with UV detection.
  • To compose the buffer along with cholic acid sodium solution (pH 9.2) for micellar electrokinetic chromatographic (MEKC) quantification of pesticides.

Legal Information

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Juan-García et al.
Journal of chromatography. A, 1073(1-2), 229-236 (2005-05-25)
Two procedures based on solid-phase extraction (SPE) and stir-bar sorptive extraction (SBSE) in combination with micellar electrokinetic chromatography (MEKC)--diode array detection (DAD) were compared for the simultaneous extraction of acrinathrin, bitertanol, cyproconazole, fludioxonil, flutriafol, myclobutanil, pyriproxyfen, and tebuconazole in lettuce
A partial modification of the carbazole method of Bitter and Muir for quantitation of hexuronic acids.
M Kosakai et al.
Analytical biochemistry, 93(2), 295-298 (1979-03-01)
The analysis of aminoglycoside antibiotics by capillary electrophoresis.
Flurer CL.
Journal of Pharmaceutical and Biomedical Analysis, 13(7), 809-816 (1995)
Crystal structures of sodium sulfate decahydrate (Glauber's salt) and sodium tetraborate decahydrate (borax). Redetermination by neutron diffraction.
Levy HA and Lisensky GC.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(12), 3502-3510 (1978)
C Cencetti et al.
Carbohydrate polymers, 90(3), 1362-1370 (2012-09-04)
Silver-loaded dressings are designed to provide the same antimicrobial activity of topical silver, with the advantages of a sustained silver release and a reduced number of dressing changes. Moreover, such type of dressing must provide a moist environment, avoiding fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service