Skip to Content
Merck
All Photos(1)

Key Documents

P1378

Sigma-Aldrich

Phthaldialdehyde

≥97% (HPLC), powder or crystals

Synonym(s):

o-Phthalaldehyde, o-Phthalic dicarboxaldehyde, Benzene-1,2-dicarboxaldehyde, OPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O2
CAS Number:
Molecular Weight:
134.13
Beilstein:
878317
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Assay

≥97% (HPLC)

form

powder or crystals

color

white to light yellow

mp

55-58 °C

storage temp.

2-8°C

SMILES string

O=Cc1ccccc1C=O

InChI

1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H

InChI key

ZWLUXSQADUDCSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Phthaldialdehyde has been used:

  • to detect Arg through spectrophotometry using end-point fluorescent sensitive detection
  • as a reagent stream with the sample to produce a fluorescent product for determining ammonia in seawater
  • in the o-phthaldialdehyde spectrophotometric assay (OPA test) to determine the proteolytic activity (PA)of the strains and the commercial cultures

Biochem/physiol Actions

o-Phthaldialdehyde (OPA) can form fluorescent derivatives while reacting with amino acids.
Phthaldialdehyde can be used for the pre-column derivatization of amino acids for high-performance liquid chromatography (HPLC) separation. It may also be used for flow cytometric measurements of protein thiol groups.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Bin Han et al.
Applied microbiology and biotechnology, 99(16), 6715-6726 (2015-02-17)
Sphingolipid ceramide N-deacylase (SCDase) catalyzes reversible reactions in which the amide linkage in glycosphingolipids is hydrolyzed or synthesized. While SCDases show great value for the enzymatic synthesis of glycosphingolipids, they are relatively poorly characterized enzymes. In this work, the enzymatic
Niklas Krause et al.
Scandinavian journal of work, environment & health, 41(2), 124-139 (2015-01-20)
This study aimed to assess the effects of physically demanding work - measured as energy expenditure (EE) during occupational physical activities (OPA) - on risk of acute myocardial infarction (AMI) among men with and without preexisting ischemic heart disease (IHD).
Bianca de Moraes Fracasso et al.
PloS one, 14(1), e0209964-e0209964 (2019-01-12)
Circulating advanced glycation end products (AGE) and their receptor, RAGE, are increased after a myocardial infarction (MI) episode and seem to be associated with worse prognosis in patients. Despite the increasing importance of these molecules in the course of cardiac
I Molnár-Perl
Journal of chromatography. A, 913(1-2), 283-302 (2001-05-18)
An overview is presented of HPLC methods currently in use to determine amino acids as their o-phthaldialdyde derivatives in the presence of various SH-group-containing additives. Crucial points that proved to influence the stability of the amino acid OPA derivatives have
M Nurul Islam et al.
The New phytologist, 193(1), 51-57 (2011-11-11)
• Sphingolipids are emerging as important mediators of cellular and developmental processes in plants, and advances in lipidomics have yielded a wealth of information on the composition of plant sphingolipidomes. Studies using Arabidopsis thaliana showed that the dihydroxy long-chain base

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service