Skip to Content
Merck
All Photos(1)

Key Documents

530050-M

Millipore

PRIMA-1

A cell-permeable quinuclidinone analog with antitumor properties.

Synonym(s):

PRIMA-1, 2,2-Bis(hydroxymethyl)-1-azabicyclo[2,2,2]octan-3-one, p53 Reactivation and Induction of Massive Apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 2 mg/mL

storage temp.

2-8°C

SMILES string

N21CCC(CC2)C(=O)C1(CO)CO

InChI

1S/C9H15NO3/c11-5-9(6-12)8(13)7-1-3-10(9)4-2-7/h7,11-12H,1-6H2

InChI key

RFBVBRVVOPAAFS-UHFFFAOYSA-N

General description

A cell-permeable quinuclidinone analog with anti-tumor properties. Induces p53-dependent apoptosis in human tumor cells through restoration of the transcriptional transactivation function to mutant p53. Restores sequence-specific DNA binding and active conformation to mutant p53 proteins in vitro and in living cells. Suppresses tumor growth in mice with no apparent toxicity. Induces apoptosis in breast cancer cells expressing mutant p53.

Biochem/physiol Actions

Primary Target
Restores biochemical and biological function to mutant p53

Warning

Toxicity: Standard Handling (A)

Other Notes

Bykov, V.J., et al. 2002. Nat. Med.8, 282.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service