Skip to Content
Merck

Skip To

F132

Fluoxetine hydrochloride

≥98% (TLC), solid, selective serotonin reuptake inhibitor 

Synonym(s):

Fluoxetine HCl, (±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
260-101-2
MDL number:
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fluoxetine hydrochloride, solid

form

solid

Quality Segment

color

white

solubility

H2O: 4 mg/mL, DMSO: >5 mg/mL

originator

Eli Lilly

SMILES string

CNCCC(C1=CC=CC=C1)OC2=CC=C(C(F)(F)F)C=C2.[H]Cl

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H

InChI key

GIYXAJPCNFJEHY-UHFFFAOYSA-N

General description

Fluoxetine hydrochloride is a psychotropic agent and one of the initial members of the anti-depressant class of drugs known as selective serotonin-reuptake inhibitors (SSRIs). It is the active ingredient of Prozac®.

Application

Fluoxetine hydrochloride has been used to study its effect on the binding ability of the radiopharmaceutical 123I-labeled 2-((2-((dimethylamino)methyl)phenyl)thio)-5-iodophenylamine ([123I]ADAM) to SERT (serotonin transporters) in mice. It has also been used for the chronic treatment of light deprived animals.It has also been used as a serotonin reuptake inhibitor to study its effects on acute stress in fish.

Biochem/physiol Actions

Fluoxetine hydrochloride works at presynaptic terminals where it prevents the reuptake of serotonin, resulting in the accumulation of serotonin in extracellular fluid at synapses.
Selective serotonin reuptake inhibitor; antidepressant.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Biogenic Amine Transporters and Chloride Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold under license from Eli Lilly and Company.
Prozac is a registered trademark of Eli Lilly and Co.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
34012PHR1394343290
description

solid

description

VETRANAL®, analytical standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

A cell-permeable phenylpropylamine derived anti-depressant that acts as a selective serotonin re-uptake inhibitor (SSRI).

form

solid

form

-

form

-

form

solid

Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

100

solubility

H2O: 4 mg/mL, DMSO: >5 mg/mL

solubility

-

solubility

-

solubility

methanol: 100 mg/mL, water: 5 mg/mL

color

white

color

-

color

-

color

-

Gene Information

human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363), SLC6A4(6532)

Gene Information

human ... SLC6A4(6532)

Gene Information

human ... SLC6A4(6532)

Gene Information

-

originator

Eli Lilly

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Fluoxetine hydrochloride


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. Good morning, can it be used for controlled release research with zein, and what would be its solubility and purity?

    1 answer
    1. Based on an online search, the use of zein in controlled release is typically for poorly water-soluble drugs. If that is the case, then this product may not be suitable for this purpose, as the solubility specification is 10 mg/mL in water, resulting in a clear solution. Purity by TLC is ≥ 98%.

      Helpful?

  2. What is the storage condition once I receive it?

    1 answer
    1. This product may be stored in room temperature.

      Helpful?

  3. What is the solubility of item F132, Fluoxetine hydrochloride? Also, how should stock solutions be stored after reconstitution?

    1 answer
    1. The solubility of Fluoxetine hydrochloride is as follows:
      • Soluble in water: 3.46 mg/mL
      •Soluble in DMSO: 34.58 ng/mL.

      After reconstitution, it is recommended to store stock solutions in tightly sealed vials at -20°C or below and to use them within 1 month.

      Helpful?

  4. Good day, I wanted to know if buying the 10 mg fluoxetine product would mean the customer would receive a pack of multiple 10 mg fluoxetine pills or if it means the product would just be 10 mg of the drug. Any information will be appreciated. Thank you.

    1 answer
    1. This product is in powder form and is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.

      Helpful?

Reviews

No rating value

Active Filters