Skip to Content

SML1789

PRIMA-1Met

≥98% (HPLC)

Synonym(s):

2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one, APR-246

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N21CCC(CC2)C(=O)C1(COC)CO

InChI

1S/C10H17NO3/c1-14-7-10(6-12)9(13)8-2-4-11(10)5-3-8/h8,12H,2-7H2,1H3

InChI key

BGBNULCRKBVAKL-UHFFFAOYSA-N

Biochem/physiol Actions

PRIMA-1Met (APR-246) is a re-activator of mutant p53 activity. It′s the methylated, more potent derivative of PRIMA-1. PRIMA-1Met covalently modifies the core domain of mutated p53 restoring the wild-type conformation and activty of p53 in tumor cells, leading to cell cycle arrest and apoptosis. PRIMA-1Met has also been shown to increase intracellular levels of reactive oxygen species (ROS), which may contribute to its anti-tumor activity.
Re-activator of mutant p53 activity
PRIMA-1Met, also called APR-246, is converted to methylene quinuclidinone (MQ), a Michael acceptor. It interacts with p53 through the cysteine (Cys) residue. It blocks the thioredoxin reductase (TrxR1) enzyme. It is under clinical investigation and well-studied. PRIMA-1Met effect is tested on breast cancer specific gene expression.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library