19-160
BrdU (5-Bromo-2′-deoxyuridine)
Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Synonym(s):
Bromodeoxyuridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
manufacturer/tradename
Upstate®
technique(s)
immunocytochemistry: suitable
shipped in
wet ice
Application
Research Category
All
All
Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine; BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Physical form
C9H11BrN2O5
Storage and Stability
2 years at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Muta. 1B - Repr. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Asian-Australasian journal of animal sciences, 27(3), 383-390 (2014-07-23)
A 15-wk feeding trial was conducted to examine the supplemental effects of Barodon on growth performance, gastrointestinal histology, feed digestibility and innate immunity in olive founder. A basal commercial diet was used as a control and two other diets were
ACS omega, 3(10), 14406-14416 (2018-11-10)
Drought is a major abiotic stress affecting crop growth and yield worldwide. Drought-induced oxidative stress results in the reduction of plant photosynthesis and reproductive success. Cerium oxide nanoparticles (nanoceria) possess potent antioxidant properties that can alleviate drought-induced oxidative stress by
Immunocytochemical detection of S-phase cells in normal and neoplastic cervical epithelium by anti-BrdU monoclonal antibody
Analytical and Quantitative Cytology and Histology, 12, 135-138 (1990)
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 186(7), 867-877 (2016-05-06)
In a process called silvering, European eels prepare for their long-distance migration from European freshwater systems to the Sargasso Sea for reproduction. During this journey, eels perform extended diel vertical migrations, and the concomitant changes in hydrostatic pressure significantly affect
The Journal of pathology, 171(2), 83-98 (1993-10-01)
Enzymatic and non-enzymatic treatments for antigen unmasking on formalin-fixed, paraffin-embedded, dewaxed sections were optimized and compared by the use of a panel of antibodies of diagnostic relevance (anti-cytokeratins, vimentin, S-100, T- and B-cell receptors, Ki-67/MIB 1, muscle actin). Non-enzymatic unmasking
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service