Skip to Content
Merck
All Photos(1)

Documents

11170376001

Roche

Anti-Bromodeoxyuridine

from mouse IgG1 (clone: BMC9318)

Synonym(s):

anti-BrdU, antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BMC9318, monoclonal

Assay

90% (HPLC and SDS-PAGE)

form

solution

packaging

pkg of 50 μg (500 μl)

manufacturer/tradename

Roche

isotype

IgG1

storage temp.

−20°C

General description

Anti-Bromodeoxyuridine is a monoclonal antibody to 5-bromo-2′-deoxyuridine-5′-monophosphate.

Specificity

The antibody specifically binds to bromodeoxyuridine and crossreacts with iodouridine (10%). Anti-bromo-deoxyuridine does not crossreact with fluorodeoxy-uridine, nor with any endogenous cellular components such as thymidine or uridine.

Immunogen

The epitope is apparently inside the DNA helix. DNA has to be denatured to ssDNA before antibody efficiently binds to DNA-BrdU.

Application

Anti-bromodeoxyuridine (Anti-BrdU) antibody is suitable for monitoring proliferating cells in blood, tissues, and tumors, as well as for determining BrdU incorporation on a single-cell level using:
  • Flow cytometry
  • Immunohistocytochemistry
  • Cryosections
  • Paraffin sections

Quality

The antibody is ≥90% pure as determined by SDS-PAGE with Coomassie-blue staining, and by HPLC.

Specifications

Preparation: BALB/c mice were immunized with a bromodeoxyuridine-bovine serum albumin conjugate. Lymphocytes isolated from the spleen were fused with Ag8.653 myeloma cells to create the BMC 9318 clone. The antibody was produced in ascites in BALB/c mice and purified by ion-exchange chromatography.
No. of tests: 250 (Flow cytometry)

Physical form

Solution, stabilized with phosphate buffered saline, pH 7.4, containing 0.09% (w/v) sodium azide and 0.2% (w/v) gelatin.

Preparation Note

Working concentration: Flow cytometry: 2 μg/ml (0.2 μg/100 μl/106 cells); Immunohistocytochemistry: 6 μg/ml
Working concentration of conjugate depends on application and substrate. Dilutions should be made in PBS (pH 7.4) containing 0.1% BSA to maintain stability of the antibody.

Analysis Note

Anti-Bromodeoxyuridine shows 10% cross reaction with iodo-deoxyuridine, but no cross reaction to fluoro-deoxyuridine.
No cross reaction to any endogenous thymidine or uridine.
Cross reactivity with 5-Br-UTP has not been tested but it is suggested that there is a good chance for reaction, because the only difference is an absent hydroxyl group on the ribose distal to the bromine substitution.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wouter Laurentius Smit et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(41), 25560-25570 (2020-09-30)
Deregulated global mRNA translation is an emerging feature of cancer cells. Oncogenic transformation in colorectal cancer (CRC) is driven by mutations in APC, KRAS, SMAD4, and TP53, known as the adenoma-carcinoma sequence (ACS). Here we introduce each of these driver
Derek C Adams et al.
American journal of physiology. Renal physiology, 297(3), F809-F815 (2009-06-19)
Long-term pulse chase experiments previously identified a sizable population of BrdU-retaining cells within the renal papilla. The origin of these cells has been unclear, and in this work we test the hypothesis that they become quiescent early during the course
Paula Duarte-Guterman et al.
Endocrinology, 160(9), 2128-2136 (2019-06-21)
Androgens (testosterone and DHT) increase adult hippocampal neurogenesis by increasing survival of new neurons in male rats and mice via an androgen receptor pathway, but it is not known whether androgens regulate neurogenesis in female rats and whether the effect
Tayyaba Jiwani et al.
Development (Cambridge, England), 147(3) (2020-01-15)
Cerebellar granule cell (GC) development relies on precise regulation of sonic hedgehog (Shh)-Gli signalling activity, failure of which is associated with motor disorders and medulloblastoma. Mutations in the pathway regulator suppressor of fused (Sufu), which modulates Gli activators and repressors
J Amigó et al.
Neuropharmacology, 111, 47-58 (2016-09-03)
Preclinical studies support a critical role of 5-HT4 receptors (5-HT4Rs) in depression and anxiety, but their influence in depression- and anxiety-like behaviours and the effects of antidepressants remain partly unknown. We evaluated 5-HT4R knockout (KO) mice in different anxiety and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service