Skip to Content
Merck
All Photos(7)

Documents

B2531

Sigma-Aldrich

Monoclonal Anti-BrdU antibody produced in mouse

clone BU-33, ascites fluid, Immunohistology Grade

Synonym(s):

Anti Brdu, Anti Brdu Antibody - Monoclonal Anti-BrdU antibody produced in mouse, Brdu Antibody, Anti-Bromodeoxyuridine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BU-33, monoclonal

species reactivity

wide range

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1000 using sections of intestine from mouse or rat treated in vivo with BrdU

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

5′-Bromo-2-deoxyuridine (BrdU) is a carcinogen that may be absorbed by skin or inhalation. BrdU solutions are sensitive to light. Monoclonal anti-BrdU (immunohistology grade) (mouse IgG1 isotype) is derived from the BU-33 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with bromodeoxyuridine conjugated to KLH.

Specificity

Reacts specifically with BrdU (bromodeoxyuridine) incorporated into DNA or coupled to a protein carrier. Cross-reaction with IdU (iododeoxyuridine) is observed.

Immunogen

bromodeoxyuridine conjugated to KLH.

Application

Monoclonal Anti-BrdU antibody has been used in immunostaining, BrdU incorporation into DNA, immunohistochemistry and uterine labeling index. It has also been used to investigate cell proliferation.

Biochem/physiol Actions

5′-Bromo-2-deoxyuridine (BrdU) staining is predominantly used to observe the multiplication of tumor cells and other tissues in vivo.

Storage and Stability

For continuous use, store at 0-5EC. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Other Notes

This product can be found as purified product that was produced using cell culture hybridoma product.
B8434 Anti-BrdU antibody, Mouse monoclonal
clone BU-33, purified from hybridoma cell culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mildronate enhances learning/memory and changes hippocampal protein expression in trained rats
Vija K, et al.
Pharmacology, Biochemistry, and Behavior, 106, 68-76 (2013)
Vascular endothelial growth factor improves functional outcome and decreases secondary degeneration in experimental spinal cord contusion injury.
Widenfalk J, et al.
Neuroscience, 120(4), 951-960 (2003)
Shuang Liang et al.
Scientific reports, 7(1), 11114-11114 (2017-09-13)
Melatonin has antioxidant and scavenger effects in the cellular antioxidant system. This research investigated the protective effects and underlying mechanisms of melatonin action in porcine somatic cell nuclear transfer (SCNT) embryos. The results suggested that the developmental competence of porcine
Dietary Lutein/Zeaxanthin Decreases Ultraviolet B-Induced Epidermal Hyperproliferation and Acute Inflammation in Hairless Mice
Gonzalez S, et al.
The Journal of Investigative Dermatology, 121(2), 399-405 (2003)
The influence of dietary isoflavone on the uterotrophic response in juvenile rats
Michael GW, et al.
Food And Chemical Toxicology, 41(11), 1517-1525 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service