Skip to Content
Merck
All Photos(1)

Key Documents

11296736001

Roche

5-Bromo-2′-deoxy-uridine Labeling and Detection Kit I

sufficient for ≤100 tests, kit of 1 (5 components), suitable for immunofluorescence

Synonym(s):

5-BrdU, 5-Bromo-2-deoxyuridine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

usage

sufficient for ≤100 tests

Quality Level

packaging

kit of 1 (5 components)

manufacturer/tradename

Roche

technique(s)

immunofluorescence: suitable

storage temp.

−20°C

General description

The kit is used for the detection of BrdU incorporated into cellular DNA using immunofluorescence microscopy. It is used for the detection of DNA synthesis by either in vitro labeling of cells or organ cultures, or by in vivo labeling, in which frozen or paraffin-embedded tissue sections must be prepared prior to fixation.
Normally, binding of the antibody is only achieved by denaturation of the DNA. This is usually obtained by exposing the cells to acid, base, or heat. These procedures result in destruction of cell integrity, including cell morphology and surface and cytoplasmatic markers.
The BrdU Labeling and Detection Kit I avoids these problems. The antibody preparation contains specific nucleases which allows access to BrdU after fixation in acidic ethanol. Therefore also simultaneous detection of other markers (double staining) is possible.

Specificity

Anti-BrdU monoclonal antibody specifically binds to 5-bromo-2′-deoxy-uridine, and shows cross-reactivity with 5-iodo-2′-deoxy-uridine (10%). Anti-BrdU shows no cross-reactivity with 5-fluoro-2′-deoxy-uridine or any endogenous cellular component, such as thymidine or uridine.

Application

Samples prelabeled with BrdU are fixed with ethanol, then incubated with a monoclonal antibody to BrdU. The antibody to BrdU supplied with the kit contains an optimized mixture of nucleases. These nucleases generate single-stranded DNA fragments, which allow binding of the antibody to BrdU without destruction of the cellular morphology. A fluorescein-labeled antibody to mouse immunoglobulin is added, then bound to the anti-BrdU antibody. Subsequently, the sample is evaluated using an immunofluorescence microscope.

  • Safe: No radioisotopes are used
  • Easy to perform: Follows a standard immunofluorescence protocol
  • Sensitive: Denaturation of DNA with nucleases allows for highly sensitive detection of BrdU
  • Flexible: Allows double-labeling protocols

BrdU Labeling and Detection Kit has been used for the detection of 5-bromo-2′-deoxy-uridine (BrdU) incorporated into cellular DNA.

Packaging

1 kit containing 5 components.

Preparation Note

Cell proliferation may be studied by monitoring the incorporation of a radioisotope, [3H]-thymidine, into cellular DNA, followed by autoradiography. Alternatively, 5-bromo-2′-deoxy-uridine (BrdU) may be used instead of thymidine. Cells that have incorporated BrdU into DNA are easily detected using a monoclonal antibody against BrdU and an enzyme- or fluorochrome-conjugated second antibody.
Working solution: BrdU labeling medium
Dilute BrdU labeling reagent 1:1000 with sterile cell culture medium (final concentration 10μM).
Note: For in vivo labeling undiluted BrdU labeling reagent (1 to 2ml/100 g body weight) is needed.
Prepare shortly before use.
Anti-BrdU working solution
Dilute anti-BrdU solution 1:10 with Incubation buffer.
Prepare shortly before use.
Anti-mouse-Ig-fluorescein stock solution
Dissolve anti-mouse-Ig-fluorescein solution in 1ml double-dist. water.
Anti-mouse-Ig-fluorescein working solution
Dilute anti-mouse Ig-fluorescein stock solution 1:10 with PBS. If an extended storage is desired, add BSA (bovine serum albumin), 10 mg/ml.
Prepare shortly before use.
Washing buffer
Dilute Washing buffer concentrate (10x) (bottle 2) 1:10 with double-dist. water.
Storage conditions (working solution): BrdU labeling medium
Store undiluted (1000x) medium in aliquots at -15 to -25°C.
Anti-BrdU working solution
Store undiluted antibody at -15 to -25°C.
Anti-mouse-Ig-fluorescein stock solution
Stable at 2 to 8°C
Washing buffer
Stable at 2 to 8°C
Sample material: Cell culture: adherent cells, suspension cells, organ, or explant cultures. Tissue sections (after in vivo labeling with BrdU).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • BrdU Labeling Reagent, sterile 1,000x concentrated

  • Washing Buffer concentrate 10x concentrated

  • Incubation Buffer

  • Anti-BrdU antibody, contains nucleases for DNA denaturation

  • Anti-mouse-Ig-fluorescein antibody

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boaz Kigel et al.
Blood, 118(15), 4285-4296 (2011-08-13)
Plexin-A4 is a receptor for sema6A and sema6B and associates with neuropilins to transduce signals of class-3 semaphorins. We observed that plexin-A1 and plexin-A4 are required simultaneously for transduction of inhibitory sema3A signals and that they form complexes. Unexpectedly, inhibition
Sophie Vasseur et al.
Oncogene, 21(11), 1685-1694 (2002-03-16)
p8 is a stress-induced DNA-binding protein, biochemically related to the architectural chromatin binding HMG protein family and whose function is presently unknown. We obtained fibroblast from mice lacking p8 and found that p8 is involved in cell growth regulation and
Nitin Sabherwal et al.
Development (Cambridge, England), 136(16), 2767-2777 (2009-07-28)
During neurogenesis in Xenopus, apicobasally polarised superficial and non-polar deep cells take up different fates: deep cells become primary neurons while superficial cells stay as progenitors. It is not known whether the proteins that affect cell polarity also affect cell
Sandra Bauer et al.
The Journal of clinical endocrinology and metabolism, 89(2), 812-822 (2004-02-07)
We have tested the hypothesis that elevated concentrations of TNF alpha could impair trophoblast invasion. Using first-trimester placental explant cultures, we have demonstrated that the cytokine inhibits in vitro migration of extravillous trophoblasts (EVT) on collagen I, and invasion through
Christoph Mück et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 65(11), 1165-1180 (2010-08-03)
Tumor necrosis factor-like cytokine 1A (TL1A) is expressed in endothelial cells and contributes to T-cell activation, via an extracellular fragment TL1A(L72-L251), generated by ectodomain shedding. Fragments of TL1A, referred to as vascular endothelial growth inhibitor, were found to induce growth

Articles

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service