Skip to Content
Merck

B9285

5-Bromo-2′-deoxyuridine

≥99%, synthetic (organic), powder

Synonym(s):

5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O5
CAS Number:
Molecular Weight:
307.10
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
200-415-9
MDL number:
Beilstein/REAXYS Number:
30395
Assay:
≥99%
Biological source:
synthetic (organic)
Form:
powder
Solubility:
NH4OH: 0.1 M at 20 °C, clear, colorless
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Bromo-2′-deoxyuridine, BioUltra, ≥99%

biological source

synthetic (organic)

Quality Segment

product line

BioUltra

assay

≥99%

form

powder

impurities

≤0.005% Phosphorus (P), ≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

191-194 °C (dec.) (lit.)

solubility

NH4OH: 0.1 M at 20 °C, clear, colorless

cation traces

Al: ≤0.0005%, Ca: ≤0.01%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.005%, Mg: ≤0.0005%, NH4+: ≤0.05%, Na: ≤0.05%, Pb: ≤0.001%, Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Br)C(=O)NC2=O

InChI

1S/C9H11BrN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

WOVKYSAHUYNSMH-RRKCRQDMSA-N

General description

5′-Bromo-2-deoxyuridine (BrdU) is a carcinogen that may be absorbed by skin or inhalation. BrdU solutions are sensitive to light.

Application

5-Bromo-2′-deoxyuridine has been used:
  • to label proliferating cells in pregnant mice in cell proliferation assay
  • to study the time course of cell proliferation at various times following ischemia, to confirm re-entry of MKI67+ cells in cell cycle to germ cells
  • in 5′-Bromo-2′Deoxyuridine (BrdUrd) staining of fibroblasts

Biochem/physiol Actions

5-Bromo-2′-deoxyuridine (5-BrdU) is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.
Thymidine analog used as a mutagen in genetic research. Selectively incorporated into cellular DNA during S-phase.
5′-Bromo-2-deoxyuridine (BrdU) staining is predominantly used to observe the multiplication of tumor cells and other tissues in vivo.


Still not finding the right product?

Explore all of our products under 5-Bromo-2′-deoxyuridine


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
B9285-250MG04061833443699
B9285-1G04061833443682
B9285-50MG04061833443705