Skip to Content
Merck
All Photos(5)

Key Documents

663131

Sigma-Aldrich

RuPhos

greener alternative

98%

Synonym(s):

2-Dicyclohexylphosphino-2′,6′-diisopropoxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43O2P
CAS Number:
Molecular Weight:
466.64
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

greener alternative product score

old score: 3
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

1-5% Bis(2′,6′-Diisopropoxybiphenyl)cyclohexylphosphine

mp

123-126 °C

functional group

phosphine

greener alternative category

SMILES string

CC(C)Oc1cccc(OC(C)C)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C30H43O2P/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25/h11-13,18-25H,5-10,14-17H2,1-4H3

InChI key

MXFYYFVVIIWKFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Design for Energy Efficiency” and “Use of Renewable Feedstock”. Click here to view its DOZN scorecard.

Application

Bulky phosphine ligand used in a palladium-catalyzed cross-coupling of aminoethyltrifluoroborates with electron-poor aryl bromides.

For small scale and high throughput uses, product is also available as ChemBeads (932205)

Legal Information

Usage subject to Patents: EP 1097158; JP 5758844; CA 2336691

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Organic letters, 9(2), 203-206 (2007-01-16)
A set of phenethylamines has been successfully prepared via Suzuki-Miyaura cross-coupling of diverse potassium beta-aminoethyltrifluoroborates with aryl halides. The potassium beta-aminoethyltrifluoroborates were easily prepared via hydroboration of enamine and enamide precursors. [reaction: see text].
Ruben Martin et al.
Accounts of chemical research, 41(11), 1461-1473 (2008-07-16)
The cores of many types of polymers, ligands, natural products, and pharmaceuticals contain biaryl or substituted aromatic structures, and efficient methods of synthesizing these structures are crucial to the work of a broad spectrum of organic chemists. Recently, Pd-catalyzed carbon-carbon
David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses
David S Surry et al.
Chemical science, 2(1), 27-50 (2011-01-01)
Dialkylbiaryl phosphines are a valuable class of ligand for Pd-catalyzed amination reactions and have been applied in a range of contexts. This review attempts to aid the reader in the selection of the best choice of reaction conditions and ligand

Articles

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

See All

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service