Merck
Dialkylbiaryl Phosphines in Pd-Catalyzed Amination: A User's Guide.
Chemical science (2011-01-01)
David S Surry, Stephen L Buchwald
ABSTRACT
Dialkylbiaryl phosphines are a valuable class of ligand for Pd-catalyzed amination reactions and have been applied in a range of contexts. This review attempts to aid the reader in the selection of the best choice of reaction conditions and ligand of this class for the most commonly encountered and practically important substrate combinations.
Sigma-Aldrich
PhDave-Phos, 97%
Sigma-Aldrich
MePhos, 97%
Sigma-Aldrich
JackiePhos, 95%
Sigma-Aldrich
BrettPhos Pd G1, Methyl t-Butyl Ether Adduct, may contain up to 1 mole equivalent of MTBE, 97%
Sigma-Aldrich
Me4tButylXphos, 96%
Sigma-Aldrich
RuPhos ChemBeads
Sigma-Aldrich
BrettPhos, 98%
Sigma-Aldrich
CyJohnPhos, 97%
Sigma-Aldrich
RuPhos, 98%
Sigma-Aldrich
tBuMePhos
Sigma-Aldrich
tBuXPhos Pd G1
Sigma-Aldrich
DavePhos, 97%
Sigma-Aldrich
RuPhos Pd G1 Methyl -Butyl Ether Adduct, 95%
Sigma-Aldrich
JohnPhos, 97%
Sigma-Aldrich
t-BuDavePhos
Sigma-Aldrich
tBuBrettPhos, 97%
Sigma-Aldrich
sSPhos
Sigma-Aldrich
SPhos, 95%
Sigma-Aldrich
SPhos, 98%
Sigma-Aldrich
SPhos Pd G1, Methyl t-Butyl Ether Adduct
Sigma-Aldrich
XPhos, 98%