Skip to Content
Merck
All Photos(8)

Key Documents

638064

Sigma-Aldrich

XPhos

greener alternative

98%

Synonym(s):

2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H49P
CAS Number:
Molecular Weight:
476.72
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-C Bond Formation

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Hiyama Coupling

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

greener alternative product score

old score: 2
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Use of Renewable Feedstocks
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

187-190 °C (lit.)

functional group

phosphine

greener alternative category

SMILES string

CC(C)C1=CC(C(C)C)=CC(C(C)C)=C1C2=C(P(C3CCCCC3)C4CCCCC4)C=CC=C2

InChI

1S/C33H49P/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3

InChI key

UGOMMVLRQDMAQQ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy”, “Use of Renewable Feedstock” and “Enhanced Catalytic Activity”. Click here to view its DOZN scorecard.
XPhos [2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl] is an air-stable electron-rich biaryl monophosphine ligand2 developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.

Application

Ligand for increased scope of Pd-catalyzed amination and amidation via arene sulfonates, aryl halides.
Ligand used in a Pd-catalyzed Suzuki coupling leading to C-15 analogs of vindoline.
Direct annulation of 2-haloanilines to indoles and tryptophans catalyzed by Pd. Synthesis of regioregular polythiophenes.
Preferred ligand for greener Sonogashira coupling in TPGS-750-M.
Preferred ligand for greener Sonogashira coupling in TPGS-750-M.

For small scale and high throughput uses, product is also available as ChemBeads (928364)

On the Way Towards Greener Transition-Metal-Catalyzed Processes as Quantified by E Factors
XPhos may be used as a ligand in the following reactions:
  • Preparation of functionalized benzylic sulfones via palladium-catalyzed Negishi cross-coupling between alkyl sulfones and aryl halides.
  • Along with pre-milled palladium(II) acetate as a pre-catalyst for the Stille cross-coupling of aryl chlorides with tributylarylstannanes to form the corresponding biaryl compounds.
  • Along with platinum chloride to catalyze the hydrosilylation of terminal arylalkynes with silanes to form functionalized β-(E)-vinylsilanes.

Legal Information

Usage subject to US Patent 7,223,879
Usage subject to US Patents 6307087 and 6395916.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 5143-5143 (2006)
Yanxing Jia et al.
The Journal of organic chemistry, 71(20), 7826-7834 (2006-09-26)
One-pot synthesis of indoles by a palladium-catalyzed annulation of ortho-haloanilines and aldehydes has been developed. Coupling of ortho-iodoaniline with aldehyde is realized under mild ligandless conditions [Pd(OAc)2, DABCO, DMF, 85 degrees C], whereas X-Phos is found to be the ligand
Palladium-Catalyzed Stille Cross-Coupling Reaction of Aryl Chlorides using a Pre-Milled Palladium Acetate and XPhos Catalyst System.
Advanced Synthesis & Catalysis, 350(7-8), 957-961 (2008)
Xphos ligand and platinum catalysts: A versatile catalyst for the synthesis of functionalized ?-(E)-vinylsilanes from terminal alkynes.
Hamze A, et al.
Journal of Organometallic Chemistry, 693(16), 2789-2797 (2008)
Peter D Johnson et al.
The Journal of organic chemistry, 71(20), 7899-7902 (2006-09-26)
Described are general protocols for the rapid construction of various C-15-substituted analogues of vindoline using palladium-catalyzed cross-coupling reactions. The required bromo- and iodovindolines were prepared in high yield by the reaction of vindoline with N-bromosuccinimide or N-iodosuccinimide, respectively. The study

Articles

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

See All

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service