Merck
Biaryl phosphane ligands in palladium-catalyzed amination.
Angewandte Chemie (International ed. in English) (2008-07-30)
David S Surry, Stephen L Buchwald
ABSTRACT
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses the application of these catalysts in C-N cross-coupling reactions in the synthesis of heterocycles and pharmaceuticals, in materials science, and in natural product synthesis.
Sigma-Aldrich
RuPhos Pd G1 Methyl -Butyl Ether Adduct, 95%
Sigma-Aldrich
t-BuDavePhos
Sigma-Aldrich
tBuBrettPhos, 97%
Sigma-Aldrich
XPhos, 98%
Sigma-Aldrich
sSPhos
Sigma-Aldrich
SPhos, 95%
Sigma-Aldrich
SPhos, 98%
Sigma-Aldrich
SPhos Pd G1, Methyl t-Butyl Ether Adduct
Sigma-Aldrich
JohnPhos, 97%
Sigma-Aldrich
PhDave-Phos, 97%
Sigma-Aldrich
MePhos, 97%
Sigma-Aldrich
JackiePhos, 95%
Sigma-Aldrich
BrettPhos Pd G1, Methyl t-Butyl Ether Adduct, may contain up to 1 mole equivalent of MTBE, 97%
Sigma-Aldrich
RuPhos, 98%
Sigma-Aldrich
RuPhos ChemBeads
Sigma-Aldrich
tBuMePhos
Sigma-Aldrich
tBuXPhos Pd G1
Sigma-Aldrich
Me4tButylXphos, 96%
Sigma-Aldrich
BrettPhos, 98%
Sigma-Aldrich
CyJohnPhos, 97%
Sigma-Aldrich
DavePhos, 97%