Skip to Content
Merck
All Photos(3)

Key Documents

638439

Sigma-Aldrich

JohnPhos

97%

Synonym(s):

(2-Biphenyl)di-tert-butylphosphine, (2-Biphenylyl)di-tert-butylphosphine, 2-(Di-tert-butylphosphino)biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4P[C(CH3)3]2
CAS Number:
Molecular Weight:
298.40
Beilstein:
8322131
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

86-88 °C (lit.)

functional group

phosphine

SMILES string

CC(C)(C)P(c1ccccc1-c2ccccc2)C(C)(C)C

InChI

1S/C20H27P/c1-19(2,3)21(20(4,5)6)18-15-11-10-14-17(18)16-12-8-7-9-13-16/h7-15H,1-6H3

InChI key

CNXMDTWQWLGCPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

JohnPhos is a Buchwald′s sterically bulky biaryl phosphine ligand. It is a reactive dialkylbiaryl phosphine ligand which catalyzes the carbon-nitrogen bond-forming reactions. Coordination chemistry of gold catalysts bearing JohnPhos as ligand has been investigated by NMR spectroscopy.

Learn more about Buchwald Phosphine Ligands

Application

Bulky biarylphosphine ligand utilized in the palladium catalyzed Stille cross-coupling reaction.
JohnPhos, a bulky phosphine ligand, was employed as catalyst in the following studies:
  • Hydrophenoxylation of unactivated internal alkynes.
  • Microwave-mediated Suzuki-Miyaura cross-coupling of benzylic bromides.
  • Pharmaceutical synthesis of novel imidazo[1,2-a]pyridines, having potent activity against the herpes virus.
  • Barluenga′s coupling of vinyl bromides with hydrazines.
  • Pd-catalyzed 2,3-diarylation of α,α-disubstituted-3-thiophenemethanols, via cleavage of C-H and C-C bonds.
Ligand utilized in amination of aryl halides and aryl triflates.

Catalyst for:
  • Decarboxylative cross-coupling of dialkoxybenzoic acids with diaryl disulfides or diaryl diselenides
  • Stereoselective preparation of imidazolidinones via intramolecular hydroamination of N-allylic-N-arylureas
  • Regioselective arylation of olefins with aryl chlorides
  • Cross-coupling reaction for the synthesis of polyunsaturated macrolactones
  • Regioselective O-alkylation reactions
  • Sonogashira-type cross coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven W McDaniel et al.
Tetrahedron letters, 52(43), 5656-5658 (2011-10-04)
A procedure for benzylic Suzuki-Miyaura cross-coupling under microwave conditions has been developed. These conditions allowed for heterocyclic compounds to be coupled. Optimum conditions found were Pd(OAc)(2), JohnPhos as the catalyst and ligand, potassium carbonate as base, and DMF as the
Artamkina, Galina A.; et al.
Synlett, 2, 235-238 (2006)
Alexander Zhdanko et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(46), 14732-14744 (2012-09-29)
Coordination chemistry of gold catalysts bearing eight different ligands [L=PPh(3), JohnPhos (L2), Xphos (L3), DTBP, IMes, IPr, dppf, S-tolBINAP (L8)] has been studied by NMR spectroscopy in solution at room temperature. Cationic or neutral mononuclear complexes LAuX (L=L2, L3, IMes
Marcia E Richard et al.
Beilstein journal of organic chemistry, 9, 2002-2008 (2013-11-10)
A range of arylgold compounds have been synthesized and investigated as single-component catalysts for the hydrophenoxylation of unactivated internal alkynes. Both carbene and phosphine-ligated compounds were screened as part of this work, and the most efficient catalysts contained either JohnPhos
Masaya Nakano et al.
The Journal of organic chemistry, 71(21), 8309-8311 (2006-10-10)
Alpha,alpha-disubstituted 3-thiophenemethanols undergo selective diarylation accompanied by cleavage of the C-H and C-C bonds of the 2- and 3-positions, respectively, upon treatment with aryl bromides in the presence of a palladium catalyst to give the corresponding 2,3-diarylthiophenes in good yields.

Articles

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

See All

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service