342890
Silicon dioxide
−325 mesh, 99.5% trace metals basis
Synonym(s):
Cristobalite, Quartz, Sand, Silica
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.5% trace metals basis
form
powder
refractive index
n20/D 1.544 (lit.)
particle size
−325 mesh
mp
1610 °C (lit.)
density
2.6 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
SiO2 is majorly used as a substrate material with excellent thermo-mechanical properties which can be used in a variety of applications which include: vapor deposition, phase deposition, atomic force microscopy probes(AFM), spin coating, electronic based devices.
Other Notes
May contain adsorbed H2O and CO2 which is removable by calcining at >900°C
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Exciton diffusion measurements in poly (3-hexylthiophene).
Shaw PE, et al.
Advanced Materials, 20(18), 3516-3520 (2008)
The effect of calcination temperature on the surface microstructure and photocatalytic activity of TiO2 thin films prepared by liquid phase deposition.
Yu J, et al.
The Journal of Physical Chemistry B, 107(50), 13871-13879 (2003)
Growth model of bamboo-shaped carbon nanotubes by thermal chemical vapor deposition.
Lee CJ and Park J
Applied Physics Letters, 77(21), 3397-3399 (2000)
Oxygen-aided synthesis of polycrystalline graphene on silicon dioxide substrates.
Chen J, et al.
Journal of the American Chemical Society, 133(44), 17548-17551 (2011)
Measurement of the adhesion force between carbon nanotubes and a silicon dioxide substrate.
Whittaker JD, et al.
Nano Letters, 6(5), 953-957 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service