Skip to Content
Merck
All Photos(2)

Key Documents

718483

Sigma-Aldrich

Silica

nanopowder, 99.8% trace metals basis

Synonym(s):

Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.8% trace metals basis

form

nanopowder

surface area

175-225 m2/g , BET

diameter

12 nm

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silica nanopowder can be used for a variety of applications such as filler material for dental applications,[1] substrate in atomic layer deposition (ALD),[2] and nanoscale insulated additive.[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of a BisGMA-resin dental restorative composites with glass, silica and titania fillers
Thorat S, et al.
Dental Materials Journal, 31(4), 635-644 (2012)
Low thermal conductivity carbon fibrous composite nanomaterial enabled by multi-scale porous structure
Gbewonyo S, et al.
Materials & Design, 134(4), 218-225 (2017)
Atomic layer deposition of stoichiometric Co3O4 films using bis (1, 4-di-iso-propyl-1, 4-diazabutadiene) cobalt
Han B, et al.
Thin Solid Films, 589(4), 718-722 (2015)
Srivalleesha Mallidi et al.
Nano letters, 9(8), 2825-2831 (2009-07-04)
Gold nanoparticles targeting epidermal growth factor receptor via antibody conjugation undergo molecular specific aggregation when they bind to receptors on cell surfaces, leading to a red shift in their plasmon resonance frequency. Capitalizing on this effect, we demonstrate the efficacy
K E Kovalenko et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1048-1060 (2013-08-24)
Boreal wetlands play an important role in global carbon balance. However, their ecosystem function is threatened by direct anthropogenic disturbance and climate change. Oil sands surface mining in the boreal regions of Western Canada denudes tracts of land of organic

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

See All

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is it porous or nonporous? I want to know particle size distribution(dispersity)

    1 answer
    1. The porosity and particle size are not determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service