Skip to Content
Merck
All Photos(1)

Key Documents

31624

Sigma-Aldrich

Silicon dioxide

acid washed and calcined, Analytical Reagent

Synonym(s):

Sand, white quartz, Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
SB.52

form

granular

quality

acid washed and calcined, Analytical Reagent

impurities

≤0.01% in HCL soluble iron (Fe)
≤0.2% soluble in HCl

loss

≤0.10% loss on ignition, 900 °C

granulation

0.1-0.3 mm, coeff var ≥96%

anion traces

chloride (Cl-): ≤50 mg/kg

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Study of silicon-silicon dioxide structure at liquid nitrogen temperature by electron spin resonance spectra has been investigated. Diffusion coefficient and solubilities of several metals (Ag, Cu, Au, Pd, and Ti) in silicon dioxide have been discussed. Microcontact printing of self-assembled monolayers (SAMs) of alkylsiloxanes on the surface of silicon dioxide has been reported. Separation and characterization of silicon dioxide (SiO2) nanostructures in dietary supplements marketed in products specifically targeted for women has been proposed.

Application

Silicon dioxide may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Wang et al.
Biosensors & bioelectronics, 18(2-3), 255-260 (2002-12-18)
A flow-injection system with an organophosphorus-hydrolase (OPH)-biosensor detector has been developed and characterized for the rapid detection of organophosphorus (OP) nerve agents. The enzyme was immobilized onto a thin-film gold detector through a cystamine-glutaraldehyde coupling. Factors influencing the performance were
Microcontact printing of octadecylsiloxane on the surface of silicon dioxide and its application in microfabrication.
Xia Y, et al.
Journal of the American Chemical Society, 117(37), 9576-9577 (1995)
Study of silicon-silicon dioxide structure by electron spin resonance I.
Nishi Y.
Jpn J. App. Phys., Part I, 10(1), 52-52 (1971)
Jin-Hee Lim et al.
Journal of agricultural and food chemistry, 63(12), 3144-3152 (2015-03-05)
Nanomaterials are beginning to enter our daily lives through various consumer products as the result of technology commercialization. The development of methodologies to detect the presence of nanomaterials in consumer products is an essential element in understanding our exposure. In
Diffusion of metals in silicon dioxide.
McBrayer JD, et al.
Journal of the Electrochemical Society, 133(6), 1242-1246 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service