Skip to Content
Merck
All Photos(1)

Key Documents

83340

Sigma-Aldrich

Sand, white quartz

purum p.a., powder

Synonym(s):

Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

form

powder

loss

≤0.3% loss on ignition, 900 °C

particle size

>230 mesh

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤200 mg/kg
K: ≤1000 mg/kg
Na: ≤150 mg/kg
Ni: ≤50 mg/kg
Pb: ≤100 mg/kg
Zn: ≤50 mg/kg

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is sand, white quartz (SiO2). Its reaction with alkaline NaNO3 solutions containing dissolved Al at 89°C has been investigated.

Application

Sand, white quartz (quartz) has been employed as a solid sample to evaluate the pore-volume variations during fluid-rock interaction experiments.

Analysis Note

metal and chloride traces refer to acid-soluble parts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pore-volume alteration measurements to evaluate scale formation during solid-fluid interactions.
Moola P, et al.
Mineralogical Magazine, 78(6), 1399-1403 (2014)
B R Bickmore et al.
Environmental science & technology, 35(22), 4481-4486 (2002-01-05)
Caustic NaNO3 solutions containing dissolved Al were reacted with quartz sand at 89 degrees C to simulate possible reactions between leaked nuclear waste and primary subsurface minerals at the U.S. Department of Energy's Hanford site in Washington. Nitrate-cancrinite began to
Mukesh K Jaiswal et al.
Journal of reproductive immunology, 112, 38-45 (2015-08-01)
The a2 isoform of vacuolar-ATPase (ATP6V0A2, referred to as a2V) is required for normal spermatogenesis and maturation of sperm. Treatment of male mice with anti-a2V disturbs the testicular cytokine/chemokine balance and leads to severe deficiencies of spermatogenesis. The aim of
G Cappelletti et al.
Environmental science and pollution research international, 22(22), 17733-17743 (2015-07-15)
In order to avoid both natural and artificial stone decay, mainly due to the interaction with atmospheric pollutants (both gases such as NOx and SO2 and particulate matter), polymeric materials have been widely studied as protective coatings enable to limit
Chang Hoon Moon et al.
Investigational new drugs, 32(3), 400-411 (2013-11-10)
CKD-516 is a benzophenone analog in which the B ring is modified by replacement with a carbonyl group. The study assessed CKD-516 as a vascular disrupting agent or anti-cancer drug. To assess the effect of S516 on vascularization, we analyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service