Skip to Content
Merck
All Photos(3)

Key Documents

262951

Sigma-Aldrich

Silicon monoxide

powder, −325 mesh

Synonym(s):

Silicon monooxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO
CAS Number:
Molecular Weight:
44.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

particle size

−325 mesh

density

2.13 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[O+]#[Si-]

InChI

1S/OSi/c1-2

InChI key

LIVNPJMFVYWSIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Silicon monoxide (SiO) is a unique compound with high optical transparency and electrical conductivity. Its ability to form thin films allows for effective use in optoelectronic devices, such as solar cells and photodetectors, where it enhances light absorption and charge transport. Additionally, SiO exhibits promising characteristics for use in battery technologies, particularly in lithium-ion batteries, where it can improve energy density and cycle stability.

Application


  • Unraveling the Fundamental Mechanism of Interface Conductive Network Influence on the Fast-Charging Performance of SiO-Based Anode for Lithium-Ion Batteries.: This paper explores the role of interface conductive networks in enhancing the fast-charging capabilities of SiO-based anodes. The research highlights the critical mechanisms influencing the performance, offering pathways to develop high-efficiency lithium-ion batteries (Zhang et al., 2023).

  • Distinctive Electric Properties of Group 14 Oxides: SiO2, SiO, and SnO2.: The study compares the electrical properties of various group 14 oxides, including silicon monoxide, to understand their potential applications in energy storage and other technologies. The findings provide a detailed analysis of SiO′s unique properties (Guerreiro et al., 2023).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jürgen Becker et al.
Nature materials, 2(1), 59-63 (2003-03-26)
In the course of miniaturization of electronic and microfluidic devices, reliable predictions of the stability of ultrathin films have a strategic role for design purposes. Consequently, efficient computational techniques that allow for a direct comparison with experiment become increasingly important.
Wei-Hsun Yeh et al.
Analytical chemistry, 83(15), 6047-6053 (2011-06-22)
Surface plasmon resonance enhanced transmission through metal-coated nanostructures represents a highly sensitive yet simple method for quantitative measurement of surface processes and is particularly useful in the development of thin film and adsorption sensors. Diffraction-induced surface plasmon excitation can produce
Taizo Kobayashi et al.
Lab on a chip, 11(4), 639-644 (2010-12-04)
This paper reports a novel combination of hydrophilic/hydrophobic materials for the evolution of liquid manipulation. Droplet generation based on a hydrophilic/hydrophobic mechanism is a promising method for highly accurate liquid manipulations. Although several droplet manipulation devices utilizing hydrophilic/hydrophobic patterns have
Jana Lokajová et al.
Electrophoresis, 31(9), 1540-1549 (2010-04-02)
Bupivacaine is a lipophilic, long-acting, amide class local anesthetic commonly used in clinical practice to provide local anesthesia during surgical procedures. Several cases of accidental overdose with cardiac arrest and death have been reported since bupivacaine was introduced to human
Arthur C Reber et al.
Nano letters, 6(6), 1190-1195 (2006-06-15)
A synergistic effort combining experiments in beams and first principles theoretical investigations is used to propose mechanisms that could lead to the formation of silicates and nanoparticles with silicon-rich cores through agglomeration of SiO, an abundant oxygen-bearing species in space.

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service