Skip to Content
Merck
All Photos(2)

Key Documents

645613

Sigma-Aldrich

Sodium oxide

80%

Synonym(s):

Disodium monoxide, Disodium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O
CAS Number:
Molecular Weight:
61.98
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

80%

form

solid

reaction suitability

core: sodium
reagent type: catalyst

impurities

~20% Na2O2

density

2.27 g/mL at 25 °C (lit.)

SMILES string

[O--].[Na+].[Na+]

InChI

1S/2Na.O/q2*+1;-2

InChI key

KKCBUQHMOMHUOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium oxide is a strong alkaline oxide, commonly used as an active flux in ceramic glazes.

Application

Sodium oxide has been used as a coating precursor on organic polymer films during the plasma modification to analyze its antimicrobial properties.
Sodium oxide has been used as a coating precursor on organic polymer films during the plasma modification to analyze its antimicrobial properties.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gianluca Malavasi et al.
The journal of physical chemistry. A, 111(34), 8401-8408 (2007-08-07)
The crystallization kinetics of Na(2)O.CaO.2SiO(2) (x = 0) and 0.68ZnO.Na(2)O.CaO.2SiO(2) (x = 0.68, where x is the ZnO stoichiometric coefficient in the glass formula) bioactive glasses have been studied using both nonisothermal and isothermal methods. The results obtained from isothermal
Gerard Klaver et al.
Environmental pollution (Barking, Essex : 1987), 148(3), 718-728 (2007-04-03)
Large hydropower dams have major impacts on flow regime, sediment transport and the characteristics of water and sediment in downstream rivers. The Gabcikovo and Iron Gate dams divide the studied Danube transect (rkm 1895-795) into three parts. In the Gabcikovo
Mathieu Digne et al.
Physical chemistry chemical physics : PCCP, 9(20), 2577-2582 (2007-05-18)
The sodium cation is well known as an efficient poison of gamma-alumina surface acidity. This poisoning effect has been revealed both by characterization methods and catalytic tests. In this work, we propose an accurate model of sodium exchanged gamma-alumina surfaces.
Qi-Zhi Chen et al.
Acta biomaterialia, 6(10), 4143-4153 (2010-05-08)
The sol-gel process of producing SiO(2)-CaO bioactive glasses is well established, but problems remain with the poor mechanical properties of the amorphous form and the bioinertness of its crystalline counterpart. These properties may be improved by incorporating Na(2)O into bioactive
Plasma deposition of antimicrobial coating on organic polymer.
Rzanek-Boroch Z, et al.
The European Physical Journal - Applied Physics, 61(2), 24316-24316 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service