Skip to Content
Merck
All Photos(2)

Key Documents

336823

Sigma-Aldrich

Silicon monoxide

pieces, 3-10 mm, 99.99% trace metals basis

Synonym(s):

Oxosilylene, Silicon oxide (SiO)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO
CAS Number:
Molecular Weight:
44.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

pieces

particle size

3-10 mm

density

2.13 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[O+]#[Si-]

InChI

1S/OSi/c1-2

InChI key

LIVNPJMFVYWSIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Grabowsky et al.
Inorganic chemistry, 48(10), 4384-4393 (2009-04-22)
Variation of a bond angle can tune the reactivity of a chemical compound. To exemplify this concept, the nature of the siloxane linkage (Si-O-Si), the most abundant chemical bond in the earth's crust, was examined using theoretical calculations on the
Elisabeth Galopin et al.
Langmuir : the ACS journal of surfaces and colloids, 26(5), 3479-3484 (2009-11-07)
The article reports on the selective adhesion of Bacillus cereus spores on patterned and heterogeneously wetted superhydrophobic silicon nanowires surfaces. Superhydrophilic patterns on superhydrophobic silicon nanowire (SiNW) surfaces were prepared by a standard optical lithography technique. Exposure of the patterned
Multiplexed protein patterns on a photosensitive hydrophilic polymer matrix.
Parijat Bhatnagar et al.
Advanced materials (Deerfield Beach, Fla.), 22(11), 1242-1246 (2010-05-04)
D Y Ju et al.
Journal of nanoscience and nanotechnology, 11(10), 8738-8743 (2012-03-10)
In this study, the composite magnetic nanoparticles of coated SiO nano film with about 8 nm size and high saturation magnetization value, were synthesized by liquid phase precipitation method. The magnetic nanoparticles can be dispersed in various liquid media, widely
Michael J R Previte et al.
Journal of fluorescence, 17(3), 279-287 (2007-04-04)
Previously we combined common practices in protein detection with chemiluminescence, microwave technology, and metal-enhanced chemiluminescence to demonstrate that we can use low power microwaves to substantially increase enzymatic chemiluminescent reaction rates on particulate silvered substrates. We now describe the applicability

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service