Merck
  • A simple, fast and cheap non-SPE screening method for antibacterial residue analysis in milk and liver using liquid chromatography-tandem mass spectrometry.
A simple, fast and cheap non-SPE screening method for antibacterial residue analysis in milk and liver using liquid chromatography-tandem mass spectrometry.
Talanta (2014-08-17)
Magda Targa Martins, Jéssica Melo, Fabiano Barreto, Rodrigo Barcellos Hoff, Louise Jank, Michele Soares Bittencourt, Juliana Bazzan Arsand, Elfrides Eva Scherman Schapoval
ABSTRACT
In routine laboratory work, screening methods for multiclass analysis can process a large number of samples in a short time. The main challenge is to develop a methodology to detect as many different classes of residues as possible, combined with speed and low cost. An efficient technique for the analysis of multiclass antibacterial residues (fluoroquinolones, tetracyclines, sulfonamides and trimethoprim) was developed based on simple, environment-friendly extraction for bovine milk, cattle and poultry liver. Acidified ethanol was used as an extracting solvent for milk samples. Liver samples were treated using EDTA-washed sand for cell disruption, methanol:water and acidified acetonitrile as extracting solvent. A total of 24 antibacterial residues were detected and confirmed using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS), at levels between 10, 25 and 50% of the maximum residue limit (MRL). For liver samples a metabolite (sulfaquinoxaline-OH) was also monitored. A validation procedure was conducted for screening purposes in accordance with European Union requirements (2002/657/EC). The detection capability (CCβ) false compliant rate was less than 5% at the lowest level for each residue. Specificity and ruggedness were also discussed. Incurred and routine samples were analyzed and the method was successfully applied. The results proved that this method can be an important tool in routine analysis, since it is very fast and reliable.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfadoxin, VETRANAL®, analytical standard
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Disodium edetate, European Pharmacopoeia (EP) Reference Standard