Skip to Content
Merck
All Photos(1)

Key Documents

17776

Millipore

Chlortetracycline Selective Supplement

suitable for microbiology

Synonym(s):

Chlortetracycline, Chlorotetracycline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClN2O8
CAS Number:
Molecular Weight:
478.88
EC Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

environmental
food and beverages

microbiology

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

suitability

Penicillium spp.

SMILES string

CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)ccc(Cl)c4C3(C)O

InChI

1S/C22H23ClN2O8/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31)/t7-,8-,15-,21-,22-/m0/s1

InChI key

CYDMQBQPVICBEU-XRNKAMNCSA-N

General description

Chemical structure: tetracycline

Application

An antibiotic supplement recommended for selective isolation of Penicillium viridicatum and related species isolated from foods.

Components

Composition (per vial; sufficient for 1000 mL medium) Chlortetracycline 50 mg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inoka D Amarakoon et al.
Journal of environmental quality, 45(5), 1644-1651 (2016-10-04)
Land application of manure containing antimicrobials results in the dispersion of the antimicrobials in agro-ecosystems. Dissipation of excreted antimicrobials in seasonally frozen agricultural soils has not been fully characterized under field conditions. This study investigated the field dissipation kinetics of
Jinsha Liu et al.
The Journal of reproduction and development, 64(2), 117-127 (2017-12-23)
Although it is known that the susceptibility of mouse spermatozoa to freezing-thawing varies greatly with genetic background, the underlying mechanisms remain to be elucidated. In this study, to map genetic regions responsible for the susceptibility of spermatozoa to freezing-thawing, we
Damian Hertig et al.
Metabolomics : Official journal of the Metabolomic Society, 15(3), 32-32 (2019-03-05)
A decline in mitochondrial function represents a key factor of a large number of inborn errors of metabolism, which lead to an extremely heterogeneous group of disorders. To gain insight into the biochemical consequences of mitochondrial dysfunction, we performed a
Ye Wang et al.
Biosensors & bioelectronics, 97, 100-106 (2017-06-05)
Novel label-free and multiplex aptasensors have been developed for simultaneous detection of several antibiotics based on a microchip electrophoresis (MCE) platform and target catalyzed hairpin assembly (CHA) for signal amplification. Kanamycin (Kana) and oxytetracycline (OTC) were employed as models for
Simona L Bavaro et al.
Frontiers in microbiology, 8, 1356-1356 (2017-08-05)
Table olives are one of the most important fermented food in the Mediterranean countries. Apart from lactic acid bacteria and yeasts that mainly conduct the olive fermentation, molds can develop on the brine surface, and can have either deleterious or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service