Skip to Content
Merck
All Photos(1)

Documents

D2900000

Disodium edetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

edta

manufacturer/tradename

EDQM

mp

248 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Voglar et al.
Environmental technology, 35(9-12), 1389-1400 (2014-04-08)
We demonstrate here, in a pilot-scale experiment, the feasibility of ethylenediaminetetraacetate (EDTA)based washing technology for soils contaminated with potentially toxic metals. Acid precipitation coupled to initial alkaline toxic metal removal and an electrochemical advanced oxidation process were used for average
Il Joong Park et al.
Archives of pathology & laboratory medicine, 138(8), 1077-1082 (2014-07-31)
The Samsung LABGEO(HC10) Hematology Analyzer (LABGEO(HC10)) is a recently developed automated hematology analyzer that uses impedance technologies. The analyzer provides 18 parameters including 3-part differential at a maximum rate of 80 samples per hour. To evaluate the performance of the
Oivind Midttun et al.
The Journal of nutrition, 144(5), 784-790 (2014-03-22)
Knowledge of stability during sample transportation and changes in biomarker concentrations within person over time are paramount for proper design and interpretation of epidemiologic studies based on a single measurement of biomarker status. Therefore, we investigated stability and intraindividual vs.
S de Vries et al.
Poultry science, 93(8), 2010-2017 (2014-06-17)
Marker methodologies to measure ileal and total tract digestibilities of diets varying in content and degradability of dietary fiber in broiler chickens were evaluated. Chromium sesquioxide (Cr2O3) and cobalt-ethylenediamine tetraacetic acid (Co-EDTA) were used as markers of solid and soluble
The role of endodontics in interdisciplinary dentistry: are you making the right decisions?
John West
Dentistry today, 33(4), 80-80 (2014-05-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service