Skip to Content
Merck
All Photos(2)

Key Documents

179957

Sigma-Aldrich

Methanol

≥99.6%, Laboratory Reagent grade

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39020101
PubChem Substance ID:
NACRES:
NA.05
grade:
Laboratory Reagent
Assay:
≥99.6%
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, Laboratory Reagent, ≥99.6%

grade

Laboratory Reagent

Quality Level

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.6%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

impurities

≤0.01% alkalinity (as NH3)
≤0.03% acidity (as HCOOH)
≤0.1% water

evapn. residue

≤0.01%

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is an industrially important alcohol.[1] Its thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported.[2] Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.[3] It participates in the purification step during the synthesis of pyocyanin, a toxic compound found in Pseudomonas aeruginosa.[4]
Methanol is an organic solvent that can be synthesized from syngas in the presence of CuO/ZnO/Al2O3 catalysts.[5] It is an ideal candidate as a hydrogen source in fuel cell technology due to its high H/C ratio, low propensity for soot generation, relatively low reforming temperature and as it exists in liquid state at room temperature.[6][7] In a direct methanol fuel cell (DMFC), methanol undergoes oxidation with air to generate electricity. The olefins (ethylene or propylene) formed from methanol via MTO (methanol-to-olefins) process, can be an alternative to oil and gas to produce hydrocarbon fuels.[8]

Application

Methanol may be used to compose the binary mixtures with acetonitrile in a study. Experimental evaluation of the values of dielectric constant and dielectric loss of these binary mixtures at microwave frequencies at 300°C has been reported.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dielectric behavior of acetonitrile+ methanol binary mixtures at microwave frequency.
Jogdand AP and Kadam PL.
IOSR Journal of Applied Physics, 2278-4861 (2014)
Standardized chemical synthesis of Pseudomonas aeruginosa pyocyanin.
Cheluvappa R.
Gastroenterologia y hepatologia, 22, 1350-1351 (2007)
Beyond oil and gas: the methanol economy.
Olah GA.
Angewandte Chemie (International Edition in English), 44(18), 2636-2639 (2005)
Catalytic carbon dioxide hydrogenation to methanol: A review of recent studies.
Jadhav SG, et al.
Chemical Engineering Research and Design, 92(11), 2557-2567 (2014)
Review of methanol reforming-Cu-based catalysts, surface reaction mechanisms, and reaction schemes.
Yong ST, et al.
International Journal of Hydrogen Energy, 38(22), 9541-9552 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service