Skip to Content
Merck
All Photos(1)

Key Documents

S6256

Sigma-Aldrich

Sulfamethazine

99.0-101.0% (on dried basis)

Synonym(s):

4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Beilstein:
261304
EC Number:
MDL number:
UNSPSC Code:
51284910
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

99.0-101.0% (on dried basis)

form

powder or crystals

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Sulfamethazine is an antibiotic used to treat bronchitis, prostatitis and urinary tract infections. It is used in disposition and depletion kinetic studies. It is used to develop detection techniques for quantification in fluids such as cows′ milk, honey and swine urine.

Biochem/physiol Actions

An antimicrobial sulfur drug. Induces CYP3A4 expression and acetylated by N-acetyltransferase. Exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11.
Sulfamethazine is an antimicrobial sulfur drug that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfamethazine is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA), which is required for bacterial synthesis of folic acid. It induces CYP3A4 expression and is acetylated by N-acetyltransferase. It exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11. Sulfamethazine is bacteriostatic.

Packaging

25G,100G

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Mitchell et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(2), 161-165 (1986-03-01)
Swine weighing 60-70 kg were orally administered 14C-sulfamethazine [4-amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene[U-14C]sulfonamide] at 12-hr intervals for 7 days (165 mg/dose; 0.126-5.04 mCi/mmol). The animals were sacrificed at 8 hr or 2, 5, or 10 days after the last dose was given and tissues
C J Chapron et al.
Journal of clinical pharmacology, 16(7), 338-344 (1976-07-01)
The relationship between sulfamethazine disposition kinetics and acetylation phenotype was studied in man. Sulfamethazine pharmacokinetic parameters were determined after the administration of the drug as an oral suspension. When the half-life, acetylation rate constant, or per cent available dose excreted
Tangbin Yang et al.
Hybridoma (2005), 29(5), 403-407 (2010-11-06)
A specific monoclonal antibody (MAb) against sulfamethazine was produced with hybridoma technology. This assay shows very high sensitivity with IC50 of 0.4 ng/mL and LOD of 0.05 ng/mL when it was run in 0.02 mol/L PBS (pH 7.5). This MAb has shown high
M Rérat et al.
Preventive veterinary medicine, 103(4), 265-273 (2011-09-29)
The present study was conducted to evaluate the efficacy of two prophylactic antibiotic treatments against bovine respiratory disease (BRD) in veal calves. In addition, the antibiotic susceptibilities of isolated Pasteurellaceae were tested. The calves were treated either on the day
Ma Jesús García-Galán et al.
The Science of the total environment, 409(24), 5505-5512 (2011-09-29)
Degradation of the sulfonamide sulfamethazine (SMZ) by the white-rot fungus Trametes versicolor was assessed. Elimination was achieved to nearly undetectable levels after 20 h in liquid medium when SMZ was added at 9 mg L(-1). Experiments with purified laccase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service