Skip to Content
Merck
All Photos(2)

Key Documents

T3258

Sigma-Aldrich

Tetracycline

98.0-102.0% (HPLC)

Synonym(s):

Tetracycline Hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · xH2O
CAS Number:
Molecular Weight:
444.43 (anhydrous basis)
Beilstein:
2230417
EC Number:
MDL number:
UNSPSC Code:
51284017
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

Assay

98.0-102.0% (HPLC)

form

powder or crystals

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

color

faint yellow to dark yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30)/t9-,10-,15-,21+,22-/m0/s1

InChI key

OFVLGDICTFRJMM-WESIUVDSSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maulik Thaker et al.
Cellular and molecular life sciences : CMLS, 67(3), 419-431 (2009-10-29)
Resistance to tetracycline emerged soon after its discovery six decades ago. Extensive clinical and non-clinical uses of this class of antibiotic over the years have combined to select for a large number of resistant determinants, collectively termed the tetracycline resistome.
Jennifer M Adams-Haduch et al.
Antimicrobial agents and chemotherapy, 52(11), 3837-3843 (2008-08-30)
A total of 49 unique clinical isolates of multidrug-resistant (MDR) Acinetobacter baumannii identified at a tertiary medical center in Pittsburgh, Pennsylvania, between August 2006 and September 2007 were studied for the genetic basis of their MDR phenotype. Approximately half of
Lucia Pallecchi et al.
Antimicrobial agents and chemotherapy, 51(8), 2720-2725 (2007-06-06)
A survey carried out in 2005 among members of a healthy population of children living in Bolivia and Peru revealed that fecal carriage of Escherichia coli strains resistant to expanded-spectrum cephalosporins was remarkably increased compared to that observed in the
Zhijie Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(13), 5004-5009 (2013-03-12)
Reported here is a piggyBac transposon-based expression system for the generation of doxycycline-inducible, stably transfected mammalian cell cultures for large-scale protein production. The system works with commonly used adherent and suspension-adapted mammalian cell lines and requires only a single transfection
Vanessa G Allen et al.
Antimicrobial agents and chemotherapy, 55(2), 703-712 (2010-11-26)
Surveillance of gonococcal antimicrobial resistance and the molecular characterization of the mechanisms underlying these resistance phenotypes are essential in order to establish correct empirical therapies, as well as to describe the emergence of new mechanisms in local bacterial populations. To

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How do I prepare a stock solution of T3258 Tetracycline? What is it soluble in?

    1 answer
    1. This product is soluble in methanol or ethanol, giving a completely clear solution at up to 30 mg/ml. The stability of this material in solution is pH-dependent. Stock solutions prepared in ethanol are estimated to be stable for 1-3 months when aliquoted and stored at -20°C. Please see the link below to review the product datasheet which also lists the solubility in 1 M HCl. Note that the potency is reduced in solutions with a pH below 2. https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/406/734/t3258dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service