Merck
  • Distribution of POPs, pesticides and antibiotic residues in organic honeys from different production areas.
Distribution of POPs, pesticides and antibiotic residues in organic honeys from different production areas.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2018-03-14)
Luca Maria Chiesa, Sara Panseri, Maria Nobile, Federica Ceriani, Francesco Arioli
ABSTRACT
Demand for honey is increasing, especially if it is organic and if its nutritional properties are linked to untreated environments in order to guarantee quality for health. Sources of contamination of honey can be divided into environmental and apicultural. Therefore, the distribution of persistent organic pollutants, pesticides and antibiotic residues from geographical areas with different contamination sources (high anthropic impact, intensive farming, husbandry and low anthropic impact) was investigated in order to confirm the potential transfer of xenobiotics into the supply chain and to give beekeepers tools for the selection of areas dedicated to organic production. The presence of polychlorinated biphenyls, polybrominated diphenyl ether and polycyclic aromatic hydrocarbons was confirmed, not only in proximity to highly urbanised centres, where the concentrations were higher, but in all environment contexts, confirming their ubiquity. No antibiotics or neonicotinoids were detected in 95 organic honeys, demonstrating the absence of apicultural treatments and consequently the good quality of honey of different areas. These results are important due to the undefined regulatory European situation on honey antibiotic limits.
Product Description
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Supelco
Fenarimol, PESTANAL®, analytical standard
Supelco
Furalaxyl, PESTANAL®, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard
Supelco
Ronidazole, analytical standard
Supelco
Quinalphos, PESTANAL®, analytical standard
Supelco
Fenamiphos, PESTANAL®, analytical standard
Supelco
Buprofezin, PESTANAL®, analytical standard
Supelco
Bupirimate, PESTANAL®, analytical standard
Supelco
Dinotefuran, PESTANAL®, analytical standard
Supelco
Florfenicol, analytical standard, for drug analysis
Supelco
Thiamethoxam, PESTANAL®, analytical standard
Supelco
Cyprodinil, PESTANAL®, analytical standard
Supelco
Ethoprophos, PESTANAL®, analytical standard
Supelco
Fludioxonil, PESTANAL®, analytical standard
Supelco
Pirimicarb, PESTANAL®, analytical standard
Supelco
Oxadixyl, PESTANAL®, analytical standard
Supelco
BDE No 154 solution, 50 μg/mL in isooctane, analytical standard
Supelco
Azoxystrobin, PESTANAL®, analytical standard
Supelco
BDE No 28 solution, 50 μg/mL in isooctane, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Cadusafos, PESTANAL®, analytical standard
Supelco
BDE No 47 solution, 50 μg/mL in isooctane, analytical standard
Supelco
BDE No 99 solution, 50 μg/mL in isooctane, analytical standard
Supelco
BDE No 153 solution, 50 μg/mL in isooctane, analytical standard
Supelco
BDE No 100 solution, 50 μg/mL in isooctane, analytical standard
Supelco
Benalaxyl, PESTANAL®, analytical standard
Supelco
Danofloxacin, VETRANAL®, analytical standard
Supelco
Florfenicol amine, VETRANAL®, analytical standard
Supelco
Ceftiofur, VETRANAL®, analytical standard