Skip to Content
Merck
All Photos(1)

Key Documents

31697

Supelco

Azoxystrobin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3O5
CAS Number:
Molecular Weight:
403.39
Colour Index Number:
23860
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CO\C=C(\C(=O)OC)c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2

InChI

1S/C22H17N3O5/c1-27-13-17(22(26)28-2)16-8-4-6-10-19(16)30-21-11-20(24-14-25-21)29-18-9-5-3-7-15(18)12-23/h3-11,13-14H,1-2H3/b17-13+

InChI key

WFDXOXNFNRHQEC-GHRIWEEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azoxystrobin belongs to strobilurin class of fungicides which is based on lead molecule strobilurin A. Its target site is mitochondrial respiration of fungi.

Application

Azoxystrobin has been used as reference for determination of azoxystrobin residue from different aquatic tox mediums using HPLC.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher C Howell et al.
Chemosphere, 112, 194-202 (2014-07-23)
The application of plant protection products has the potential to significantly affect soil microbial community structure and function. However, the extent to which soil microbial communities from different trophic levels exhibit resistance and resilience to such compounds remains poorly understood.
Elsa Teresa Rodrigues et al.
Environment international, 53, 18-28 (2013-01-15)
The use of pesticides for crop protection may result in the presence of toxic residues in environmental matrices. In the aquatic environment, pesticides might freely dissolve in the water or bind to suspended matter and to the sediments, and might
Xin Guo et al.
Electrophoresis, 38(16), 2004-2010 (2017-05-13)
A nonaqueous micellar electrokinetic capillary chromatography method with indirect LIF was developed for the determination of strobilurin fungicide residues in fruits and vegetables. Hydrophobic CdTe quantum dots (QDs) synthesized in aqueous phase were used as background fluorescent substance. The BGE
Yingnan Han et al.
Ecotoxicology and environmental safety, 107, 214-219 (2014-07-11)
Azoxystrobin has been widely used in recent years. The present study investigated the oxidative stress and DNA damage effects of azoxystrobin on earthworms (Eisenia fetida). Earthworms were exposed to different azoxystrobin concentrations in an artificial soil (0, 0.1, 1, and
Majid Ali Khan et al.
The Science of the total environment, 573, 1573-1579 (2016-09-20)
Studies with small soil columns (2cm i.d.×5.4cm depth) compared leaching of four pesticides added either as technical material or as commercial formulations. Pesticides were selected to give a gradient of solubility in water between 7 and 93mgL-1, comprising azoxystrobin (emulsifiable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service