Skip to Content
Merck
All Photos(1)

Key Documents

45627

Supelco

Pirimicarb

PESTANAL®, analytical standard

Synonym(s):

2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N4O2
CAS Number:
Molecular Weight:
238.29
Beilstein:
663442
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C(=O)Oc1nc(nc(C)c1C)N(C)C

InChI

1S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3

InChI key

YFGYUFNIOHWBOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fabienne Dulin et al.
Parasitology research, 113(12), 4601-4610 (2014-11-02)
Varroa destructor is the main concern related to the gradual decline of honeybees. Nowadays, among the various acaricides used in the control of V. destructor, most presents increasing resistance. An interesting alternative could be the identification of existent molecules as
Andrea X Silva et al.
PloS one, 7(6), e36366-e36366 (2012-06-12)
Insecticide resistance is one of the best examples of rapid micro-evolution found in nature. Since the development of the first synthetic insecticide in 1939, humans have invested considerable effort to stay ahead of resistance phenotypes that repeatedly develop in insects.
Guowen Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 687-694 (2010-12-24)
The interaction between pirimicarb and calf thymus DNA in physiological buffer (pH 7.4) was investigated with the use of Neutral Red (NR) dye as a spectral probe by UV-vis absorption, fluorescence and circular dichroism (CD) spectroscopy, as well as viscosity
Frank Seitz et al.
Environmental toxicology and chemistry, 31(3), 518-523 (2011-12-16)
Titanium dioxide nanoparticles (nTiO₂) form reactive oxygen species (ROS) under irradiation by ultraviolet light (UV). This known photocatalytic activity may finally affect the presence and toxicity of organic environmental chemicals, which have not yet been studied at ambient UV intensity.
Encarnación Rodríguez-Gonzalo et al.
Electrophoresis, 29(19), 4066-4077 (2008-10-30)
CZE was assayed for the separation of carbamate pesticides susceptible to protonation (Pirimicarb, Carbendazim). Different electrophoretic media with high organic contents were explored, adequate separation and resolution being achieved when a BGE based on ACN with acetic acid in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service