Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
COCC(=O)N(N1CCOC1=O)c2c(C)cccc2C
InChI
1S/C14H18N2O4/c1-10-5-4-6-11(2)13(10)16(12(17)9-19-3)15-7-8-20-14(15)18/h4-6H,7-9H2,1-3H3
InChI key
UWVQIROCRJWDKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Oxadixyl is a fungicide used primarily for the control of late blight disease in crops.
Application
Oxadixyl may be used as a reference standard for the determination of the analyte in vegetables, rice, fresh foodstuffs and dry animal feed by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fast and sensitive determination of pesticide residues in vegetables using low-pressure gas chromatography with a triple quadrupole mass spectrometer.
Journal of Chromatography A, 1128(1-2), 236-243 (2006)
Determination of 81 multiclass pesticides in fresh foodstuffs by a single injection analysis using gas chromatography-chemical ionization and electron ionization tandem mass spectrometry.
Analytica Chimica Acta, 484(2), 167-180 (2003)
Communications in agricultural and applied biological sciences, 76(4), 909-914 (2011-01-01)
Lake Tota (Boyaca, Colombia) supplies water for human consumption, agriculture and industry for more than 500.000 people. Oxadixyl and Tebuconazole are fungicides used in onion crops in the lake catchment area. The mobility of pesticides in soil, bioavailability and transfer
Disease development and buildup of resistance to oxadixyl in potato crops inoculated with Phytophthora infestans as affected by oxadixyl and oxadixyl mixtures: Experimental and simulation studies.
Journal of Phytopathology, 132(3), 219-229 (1991)
[The establishment of the TLV of the fungicide sandofan in soil].
Gigiena i sanitariia, (1)(1), 28-28 (1993-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service