Skip to Content
Merck
All Photos(2)

Key Documents

32505

Supelco

Cadusafos

PESTANAL®, analytical standard

Synonym(s):

O-Ethyl S,S-bis(1-methylpropyl) phosphorodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H23O2PS2
CAS Number:
Molecular Weight:
270.39
Beilstein:
8140049
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(SC(C)CC)SC(C)CC

InChI

1S/C10H23O2PS2/c1-6-9(4)14-13(11,12-8-3)15-10(5)7-2/h9-10H,6-8H2,1-5H3

InChI key

KXRPCFINVWWFHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cadusafos is a nematicide used in controlling nematodes, soil insects and cutworms in a variety of fruits and vegetables. Its mode of action involves the inhibition of acetylcholinesterase.

Application

Cadusafos may be used as a reference standard in the determination of cadusafos in Radix astragali plant using microwave-assisted extraction (MAE) and dispersive solid-phase extraction (D-SFE) with gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

264.9 °F - closed cup

Flash Point(C)

129.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Simultaneous determination of organophosphorus, organochlorine, pyrethriod and carbamate pesticides in Radix astragali by microwave-assisted extraction/dispersive-solid phase extraction coupled with GC--MS
Mao X, et al.
Talanta, 97(1), 131-141 (2012)
Metabolic Pathways of Agrochemicals: Insecticides and fungicides (1998)
Mobility and dissipation of cadusafos in banana fields in Martinique
Zheng.QS, et al.
The Science of the Total Environment, 156(1), 1-9 (1994)
Dao Feng Xiang et al.
Biochemistry, 54(51), 7539-7549 (2015-12-03)
The most familiar organophosphorus compounds are the neurotoxic insecticides and nerve agents. A related group of organophosphorus compounds, the phosphotriester plasticizers and flame retardants, has recently become widely used. Unlike the neurotoxic phosphotriesters, the plasticizers and flame retardants lack an
Mario Masís-Mora et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1156, 122296-122296 (2020-08-24)
Biopurification systems are useful in the management of pesticide residues and provide an option to dispose wastewaters of agricultural origin derived from pesticide application practices. The analysis of pesticide residues in the biopurification system biomixture is necessary to determine whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service