Skip to Content
Merck
All Photos(3)

Documents

Q1250

Sigma-Aldrich

Quinine hemisulfate salt monohydrate

synthetic, ≥90% (HPLC)

Synonym(s):

Quinine sulfate (2:1) (salt) dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47
Beilstein:
6113937
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥90% (HPLC)

mp

~225 °C (dec.) (lit.)

solubility

H2O: slightly soluble 1.2 mg/mL
ethanol: 8 mg/mL

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

originator

Bayer

SMILES string

O.O.OS(O)(=O)=O.COc1ccc2nccc([C@@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)C7CC8CCN7C[C@@H]8C=C)c6c5

InChI

1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1

InChI key

ZHNFLHYOFXQIOW-LPYZJUEESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Quinine hemisulfate salt monohydrate has been used as a stimuli to evaluate its influence on sensory and cognitive factors.

Biochem/physiol Actions

Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive, and hypoglycemic agent; alkaloid originally isolated from the Cinchona family of South American trees. Inhibits mitochondrial ATP-regulated potassium channel. Used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
Quinine has analgesic property.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A Sneddon et al.
Alcoholism, clinical and experimental research, 44(7), 1400-1409 (2020-05-31)
More women are being diagnosed with alcohol use disorder (AUD), are increasing the amount of alcohol they are drinking, and are partaking in risky drinking behaviors. Compulsive drinking which persists despite negative consequences is a hallmark of AUD. Preclinical aversion-resistant
Elizabeth A Sneddon et al.
Alcoholism, clinical and experimental research, 43(2), 243-249 (2018-11-16)
Alcohol use disorder is characterized by compulsive alcohol intake, or drinking despite negative consequences. Previous studies have shown that female rodents have a heightened vulnerability to drug use across different stages of the addictive cycle, but no previous studies have
Both perceptual and conceptual factors influence taste-odor and taste-taste interactions
Frank RA, et al.
Perception & Psychophysics, 54(3), 343-354 (1993)
A Hedman et al.
Journal of pharmaceutical sciences, 87(4), 457-461 (1998-04-21)
The pharmacokinetic interaction between quinidine and digoxin in patients is well-known, in general requiring a dose reduction of digoxin in patients concomitantly treated with quinidine. Quinine, the diastereomer of quinidine, has not been as extensively studied in this respect. In
E M Clement et al.
Neuropharmacology, 37(7), 945-951 (1998-10-17)
Quinine and quinidine are reported to potentiate the behavioural effects of serotonergic agents and monoamine uptake inhibitors. We have therefore investigated the presynaptic actions of quinine and quinidine on monoamine uptake and release in rat brain tissue in vitro. Quinidine

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service