Skip to Content
Merck
All Photos(1)

Key Documents

22632

Sigma-Aldrich

Quinine hydrochloride dihydrate

tested according to Ph. Eur.

Synonym(s):

Chinini hydrochloridum, (8α, 9R)-6′-Methoxycinchonan-9-ol monohydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · 2H2O
CAS Number:
Molecular Weight:
396.91
Beilstein:
6112655
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.25

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

solid

solubility

water: soluble 0.25 mg in 5 ml

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.Cl.COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2.ClH.2H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;2*1H2/t13-,14-,19-,20+;;;/m0.../s1

InChI key

MPQKYZPYCSTMEI-FLZPLBAKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Quinine hydrochloride dehydrate has been used for fluorescent in situ detection of quinine in cinchona bark using UV resonance raman spectroscopy. It has also been used as a actinometric system to measure the light-induced changes in ruthenium anticancer agent, NAMI-A.

Biochem/physiol Actions

Quinine is a very effective anti-malarial treatment but is considered toxic for routine treatment due to the development of drug resistance in the malarial parasites. It acts as blood schizonticide.Quinine reportedly blocks the membrane conductance of potassium in enteric neurons.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marjan Bouma et al.
Journal of pharmaceutical and biomedical analysis, 30(4), 1287-1296 (2002-11-01)
NAMI-A is a novel ruthenium complex with selective activity against metastases currently in Phase I clinical trials in The Netherlands. The photostability of this new agent in solid state and in solution has been investigated utilizing a stability-indicating reversed-phase high
Torsten Frosch et al.
The journal of physical chemistry. B, 111(16), 4171-4177 (2007-03-31)
Deep UV resonance Raman micro-spectroscopy (lambda(exc) = 244 nm) was applied for a highly sensitive, selective, and gentle localization of the antimalarial quinine in situ in cinchona bark. The high potential of the method was demonstrated by the detection of
Is alkylation the main mechanism of action of the antimalarial drug artemisinin?.
Robert A and Meunier B
Chemical Society Reviews, 273-274 (1998)
E Cherubini et al.
British journal of pharmacology, 83(1), 3-5 (1984-09-01)
Quinine (100 microM) abolished the slow calcium-dependent afterhyperpolarization which occurs after an action potential in some neurones of the guinea-pig myenteric and submucous plexus. This occurred without any effect on the amplitude or time course of the action potential itself
D A Warrell
Journal of the Royal Society of Medicine, 82 Suppl 17, 44-50 (1989-01-01)
In the treatment of severe Plasmodium falciparum infection antimalarial drugs should, ideally, be given by controlled rate intravenous infusion until the patient is able to swallow tablets. In cases where infection has been acquired in a chloroquine resistant area, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service