Skip to Content
Merck
All Photos(3)

Key Documents

Q0875

Sigma-Aldrich

Quinidine sulfate salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H48N4O4 · H2O4S · 2H2O
CAS Number:
Molecular Weight:
782.94
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

impurities

≤20% Dihydroquinidine (according to USP specifications., actual content given on label)

mp

212-214 °C (dec.) (lit.)

originator

Bayer

SMILES string

[H]O[H].[H]O[H].OS(O)(=O)=O.[H][C@]1(CN2CCC1C[C@]2([H])[C@@H](O)c3ccnc4ccc(OC)cc34)C=C.[H][C@]5(CN6CCC5C[C@]6([H])[C@@H](O)c7ccnc8ccc(OC)cc78)C=C

InChI

1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14?,19-,20+;;;/m11.../s1

InChI key

ZHNFLHYOFXQIOW-WFTMRWCJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinidine is an alkaloid present in the bark of the Cinchona tree. It is the D-isomer of quinine, an antimalarial drug.

Application

Quinidine sulfate salt dihydrate has been used as an antiarrhythmic drug to study its effects on phosphorylation and mutations in the cardiac sodium channel Nav1.5.

Biochem/physiol Actions

Class IA antiarrhythmic; potassium channel blocker.
Quinidine shows therapeutic effects against atrial fibrillation and cardiac arrhythmias.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Vicente et al.
Journal of the American Heart Association, 4(4) (2015-04-15)
Congenital long QT syndrome type 2 (abnormal hERG potassium channel) patients can develop flat, asymmetric, and notched T waves. Similar observations have been made with a limited number of hERG-blocking drugs. However, it is not known how additional calcium or
Motoya Suzuki et al.
Xenobiotica; the fate of foreign compounds in biological systems, 44(11), 1039-1045 (2014-05-21)
1.  This study was aimed to characterize gastrointestinal absorption of digoxin using wild-type (WT) and multidrug resistance protein 1a [mdr1a; P-glycoprotein (P-gp)] knockout (-/-) rats. 2.  In WT rats, the area under the plasma concentration-time curve (AUC) of oral digoxin
Yuki Ogawa et al.
European journal of pediatrics, 174(4), 509-518 (2014-09-25)
This study aimed to determine the population pharmacokinetics of doxapram in low-birth-weight (LBW) infants. A total of 92 serum concentration measurements that were obtained from 34 Japanese neonates were analyzed using nonlinear mixed-effect modeling (NONMEM). Estimates generated by NONMEM indicated
Maria Addolorata Saracino et al.
Journal of pharmaceutical and biomedical analysis, 95, 61-67 (2014-03-19)
A rapid and reliable analytical method has been developed to quantify the melatonergic antidepressant agomelatine in three matrices, and namely saliva, plasma and dried blood spots. The method is based on the use of liquid chromatography with fluorimetric detection exploiting
Aleksandra Kasperczyk et al.
Magnesium research, 28(1), 14-22 (2015-05-15)
The present study investigated associations between environmental exposure to magnesium (Mg) and the levels of oxidative stress parameters and selected cytokines, and the antioxidant defense system in the seminal plasma of fertile males. The study population consisted of 57 healthy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service