Skip to Content
Merck
All Photos(3)

Key Documents

G1128

Sigma-Aldrich

Tragacanth

Synonym(s):

Gum tragacanth

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant (Astragalus gummifer)

Quality Level

form

powder

color

beige

solubility

H2O: 20 mg/mL

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Tragacanth is a gum commonly found in the sap of various Middle Eastern legumes. It is commonly used as freezing-embedding medium for tissue preservation.

Application

When mixed with water, tragacanth yields a colloidal hydrosol. The bassorin fraction (which consists of approx. 60-70% of the compound) swells to form a gel. This swelling gives the gum tragacanth the ability to form thick, viscous dispersions and pastes. It is an effictive emulsion stabilizer.

Other Notes

A complex mixture of polysaccharides including tragacanthin and bassorin.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad Amin Mohammadifar et al.
International journal of biological macromolecules, 38(1), 31-39 (2006-02-14)
Solution properties of tragacanthin (the water-soluble part of gum tragacanth) were studied by gel permeation chromatography (GPC) combined with multi-angle light scattering and viscometry at 25 degrees C. Photon correlation spectroscopy was used to determine the hydrodynamic radius. Ultrasonic degradation
Masoud Hatami et al.
International journal of biological macromolecules, 50(4), 1109-1115 (2012-03-13)
The aim of this study was to investigate the role of process conditions and system composition on the acid-induced gelation of a mixture of milk protein and gum tragacanth. This was studied by determining the effects of co-solute (lactose) addition
A Shimotoyodome et al.
Biofouling, 22(3-4), 261-268 (2007-02-13)
The aim of this study was to investigate materials which reduce saliva-promoted adhesion of Streptococcus mutans onto enamel surfaces, and their potential in preventing dental biofilm development. The effects of hydroxyapatite (HA) surface pretreatment with hydrophilic polysaccharides on saliva-promoted S.
A Hagiwara et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(8), 673-679 (1992-08-01)
Tragacanth gum was administered at dietary levels of 0 (control), 1.25 and 5.0% to groups of 50 male and 50 female B6C3F1 mice for 96 wk after which all animals were maintained on a basal diet without tragacanth gum for
T M Wieland et al.
Journal of animal science, 71(7), 1863-1868 (1993-07-01)
Four trials were conducted using 86, 24-h-old pigs to evaluate the utilization of medium-chain triglycerides (MCT). Effects of emulsification and amount of MCT fed were examined. After a 4-h period during which feed was withheld, pigs were force-fed MCT (containing

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the average molecular weight?

    1 answer
    1. This material is comprised of heterogeneous polysaccharides. The molecular weight has not been determined. Various literature sources suggest a molecular weight of approximately 840 kDa. Please see the link below to review additional information:
      https://www.sciencedirect.com/topics/medicine-and-dentistry/gum-tragacanth#:~:text=Tragacanth%20gum%20is%20a%20plant,et%20al.%2C%202003).

      Note that PubChem indicates a molecular weight of 150.35 g/mol for the monomer as indicated at the link below:
      https://pubchem.ncbi.nlm.nih.gov/compound/Gum-tragacanth

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service