Skip to Content
Merck
All Photos(2)

Key Documents

69311

Supelco

Quinine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

6′-Methoxycinchonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Beilstein:
91867
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

173-175 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1

InChI key

LOUPRKONTZGTKE-WZBLMQSHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potassium channel blocker

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Store under argon.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joop E Arends et al.
The American journal of tropical medicine and hygiene, 89(3), 527-530 (2013-07-17)
Recently, two patients of African origin were given a diagnosis of Plasmodium falciparum malaria without recent travel to a malaria-endemic country. This observation highlights the importance for clinicians to consider tropical malaria in patients with fever. Possible transmission routes of
André Felipe Cândido da Silva et al.
Medical history, 58(1), 1-26 (2013-12-18)
This article addresses the discussion about quinine-resistant malaria plasmodium in the early decades of the twentieth century. Observed by Arthur Neiva in Rio de Janeiro in 1907, the biological and social resistance of malaria sufferers to preventive and curative treatment
Annemarie R Kreeftmeijer-Vegter et al.
Malaria journal, 12, 115-115 (2013-03-30)
Exchange transfusion (ET) has remained a controversial adjunct therapy for the treatment of severe malaria. In order to assess the relative contribution of ET to parasite clearance in severe malaria, all patients receiving ET as an adjunct treatment to parenteral
Jane Achan et al.
Malaria journal, 10, 144-144 (2011-05-26)
Quinine remains an important anti-malarial drug almost 400 years after its effectiveness was first documented. However, its continued use is challenged by its poor tolerability, poor compliance with complex dosing regimens, and the availability of more efficacious anti-malarial drugs. This
Hmwe Hmwe Kyu et al.
Bulletin of the World Health Organization, 87(12), 896-904 (2010-05-11)
To summarize the existing evidence on the efficacy of artemether and arteether, two artemisinin derivatives, versus quinine for treating cerebral malaria in children. We searched the Cochrane Central Register of Controlled Trials, MEDLINE, EMBASE and the http://clinicaltrials.gov web site. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service