Skip to Content
Merck
All Photos(1)

Documents

P8396

Sigma-Aldrich

Piperacillin sodium salt

penicillin analog

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N5NaO7S
CAS Number:
Molecular Weight:
539.54
Beilstein:
5373920
EC Number:
MDL number:
UNSPSC Code:
51282423
PubChem Substance ID:
NACRES:
NA.85

form

powder

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)c4ccccc4)C(=O)C1=O

InChI

1S/C23H27N5O7S.Na/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28;/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34);/q;+1/p-1/t13-,14-,15+,20-;/m1./s1

InChI key

WCMIIGXFCMNQDS-IDYPWDAWSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Piperacillin is a semisynthetic, broad-spectrum ureidopenicillin antibiotic. It is derived from ampicillin. It has been used in pharmacokinetic studies in order to optimize antimicrobial therapy in patients with sepsis. It is used to study piperacillin hypersensitivity reactions and to study multidrug-resistant organisms.

Biochem/physiol Actions

Piperacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Piperacillin may interfere with autolysin inhibitors.

Packaging

1g,5g,10g

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Whitaker et al.
Journal of immunology (Baltimore, Md. : 1950), 187(1), 200-211 (2011-05-25)
A mechanistic understanding of the relationship between the chemistry of drug Ag formation and immune function is lacking. Thus, mass spectrometric methods were employed to detect and fully characterize circulating Ags derived from piperacillin in patients undergoing therapy and the
Saloni Singla et al.
The Journal of antibiotics, 66(2), 61-66 (2012-11-22)
The existence of majority of bacteria in biofilm mode makes it difficult to eradicate them as antibiotics at much higher concentrations than the MICs are required to destroy these bacteria. This study investigated the effect of different classes of antibiotics
Sadikah Behbehani et al.
Obstetrics and gynecology, 121(2 Pt 2 Suppl 1), 447-449 (2013-02-01)
Oxidized regenerated cellulose is a topical hemostatic agent that is used commonly in abdominal and pelvic surgery. Although oxidized regenerated cellulose mimicking an abscess has been reported after different operations, little is known about its side effects after gynecologic surgery.
Grace C Lee et al.
Clinical therapeutics, 34(12), 2297-2300 (2012-12-01)
Studies evaluating the outcomes of an extended-infusion (EI) piperacillin-tazobactam dosing strategy in specific cohorts of critically ill patients are lacking. A retrospective, pre-implementation and post-implementation study of 148 critically ill patients was conducted to compare EI and traditional infusion piperacillin-tazobactam.
Zhiping Li et al.
European journal of clinical pharmacology, 69(6), 1223-1233 (2013-01-29)
To develop population pharmacokinetic (PK) models for piperacillin/tazobactam in neonates and infants of less than 2 months of age in order to determine the appropriate dosing regimen and provide a rational basis for the development of preliminary dosing guidelines suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service