Skip to Content
Merck
All Photos(1)

Documents

C3809

Sigma-Aldrich

Ceftazidime hydrate

90.0-105.0%, contains ~10% sodium carbonate as stabilizer

Synonym(s):

1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N6O7S2 · xH2O
CAS Number:
Molecular Weight:
546.58 (anhydrous basis)
MDL number:
UNSPSC Code:
51284108
PubChem Substance ID:
NACRES:
NA.85

Assay

90.0-105.0%

form

powder or crystals

contains

~10% sodium carbonate as stabilizer

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(C)(O\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[n+]3ccccc3)=C(N2C1=O)C([O-])=O)c4csc(N)n4)C(O)=O

InChI

1S/C22H22N6O7S2.5H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;;;;;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);5*1H2/b26-13-;;;;;/t14-,18-;;;;;/m1...../s1

InChI key

NMVPEQXCMGEDNH-TZVUEUGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Broad spectrum antibiotic; third generation cephalosporin
Chemical structure: ß-lactam

Application

Ceftazidime hydrate is used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP3 on bacterial cell wall mucopeptide synthesis. It is also used in bacterial susceptibility studies and to select mutant colonies of interest.

Biochem/physiol Actions

Cephalosporins, such as ceftazidime , disrupt the synthesis of the peptidoglycan layer of bacterial cell walls.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neang S Ly et al.
The Journal of antimicrobial chemotherapy, 70(5), 1434-1442 (2015-02-26)
Colistin is an 'old' drug, which is being increasingly utilized due to limited therapeutic options. However, resistance emergence during monotherapy is concerning. Here, our objective was to optimize colistin combinations against Pseudomonas aeruginosa by profiling the time course of synergistic
Melissa D Barnes et al.
mBio, 9(6) (2018-12-13)
Pseudomonas aeruginosa produces a class C β-lactamase (e.g., PDC-3) that robustly hydrolyzes early generation cephalosporins often at the diffusion limit; therefore, bacteria possessing these β-lactamases are resistant to many β-lactam antibiotics. In response to this significant clinical threat, ceftolozane, a
Kristopher R Genschmer et al.
Cell, 176(1-2), 113-126 (2019-01-12)
Here, we describe a novel pathogenic entity, the activated PMN (polymorphonuclear leukocyte, i.e., neutrophil)-derived exosome. These CD63+/CD66b+ nanovesicles acquire surface-bound neutrophil elastase (NE) during PMN degranulation, NE being oriented in a configuration resistant to α1-antitrypsin (α1AT). These exosomes bind and
Borys Snopok et al.
The Analyst, 140(9), 3225-3232 (2015-03-13)
An ultra-sensitive gas phase biosensor/tracer/bio-sniffer is an emerging technology platform designed to provide real-time information on air-borne analytes, or those in liquids, through classical headspace analysis. The desired bio-sniffer measures gaseous 17α- ethinylestradiol (ETED) as frequency changes on a quartz
Ruud R G Bueters et al.
Pediatric research, 76(6), 508-514 (2014-08-29)
Many premature born neonates receive antibiotic drugs to treat infections, which are applied during active nephrogenesis. We studied the impact of clinical concentrations of gentamicin and alternatives, ceftazidime and meropenem, on ureteric branching. Mice metanephroi were dissected at embryonic day

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service