- Outcomes of extended-infusion piperacillin-tazobactam: a retrospective analysis of critically ill patients.
Outcomes of extended-infusion piperacillin-tazobactam: a retrospective analysis of critically ill patients.
Clinical therapeutics (2012-12-01)
Grace C Lee, Hansheng Liou, Russell Yee, Clifford F Quan, Katherine Neldner
PMID23195962
ABSTRACT
Studies evaluating the outcomes of an extended-infusion (EI) piperacillin-tazobactam dosing strategy in specific cohorts of critically ill patients are lacking. A retrospective, pre-implementation and post-implementation study of 148 critically ill patients was conducted to compare EI and traditional infusion piperacillin-tazobactam. In this retrospective study, the EI piperacillin-tazobactam dosing strategy was associated with improved 30-day mortality. EI piperacillin-tazobactam may be an effective alternative to TI of piperacillin-tazobactam among critically ill patients treated for suspected gram-negative infections.
MATERIALS