This product is not tested for moisture content.
17850
Ciprofloxacin
≥98% (HPLC)
Synonym(s):
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, Ciprobay
About This Item
Recommended Products
Agency
EPA 1694
Quality Level
Assay
≥98% (HPLC)
form
powder or crystals
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
application(s)
environmental
Mode of action
DNA synthesis | interferes
enzyme | inhibits
SMILES string
OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O
InChI
1S/C17H18FN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)
InChI key
MYSWGUAQZAJSOK-UHFFFAOYSA-N
Gene Information
human ... CYP1A2(1544) , KCNH1(3756)
rat ... Gabra1(29705)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Chemical structure: fluoroquinolone
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
is this the anhydrous form of ciprofloxacin?
1 answer-
Helpful?
-
-
Is Ciprofloxacin HCL available and is it USP grade?
1 answer-
Product 17850 has a molecular formula of C17H18FN3O3. The form of the chemical being searched for would have a molecular formula of C17H18FN3O3.HCL,H2O. There is Ciprofloxacin HCL.H2O but only in very small quantities as a secondary pharmaceutical standard, with the catalog number PHR1044. Regarding the question about whether the products are USP grade, the answer is no. Both products are labeled as strictly for Research Use Only and, as such, are not suitable for human use.
Helpful?
-
-
Hi. Should I keep ciprofloxacin 17850-5g in refrigerator or -20? Thank you
1 answer-
In the powder form, this product is to be stored at tightly closed at room temperature. Solutions prepared in 1% HCl at 1 mg/ml may be stored in frozen aliquots at -20 °C for up to 12 months.
Helpful?
-
-
What are the storage conditions of this product?
1 answer-
This product is to be stored at room temperature.
Helpful?
-
-
17850 Sigma-Aldrich Ciprofloxacin How much is the measured water content and active fraction of the above product?
1 answer-
This product is not tested for moisture content or potency/activity. The minimum purity of this product is 98% by HPLC. A similar product, PHR1167, is tested for loss on drying, but not potency/activity. This product typically demonstrates a >99% purity as compared to the USP and EP standards. See the link below to review this product option:
https://www.sigmaaldrich.com/US/en/product/sial/phr1167Helpful?
-
-
If a stock solution of Cipro 17850 is solved in 0,1 HCl, at what temperature should it be stored and how long ist it stable?
1 answer-
Unfortunately, stability studies have not been performed on this product. However, several papers have information on the solution stability of ciprofloxacin.
(1) Stock solutions of ciprofloxacin in water at 200 mg/L, 100 mg/L, 40 mg/L, 20 mg/L and 4 mg/L can be stored at -20 °C for "at least 60 days.
http://www.clinchem.org/cgi/reprint/31/5/684.pdf
(2) Stock solutions of ciprofloxacin at 1 mg/mL in 1% HCl can be stored at -20 °C for at least 12 months.
http://onlinelibrary.wiley.com/doi/10.1002/bmc.1523/fullHelpful?
-
-
Is ciprofloxacin 17850 soulable in water
1 answer-
This product is soluble in DMSO (<1 mg/ml at 25° C), methanol, 0.1N hydrochloric acid (25 mg/ml), and ethanol (<1 mg/ml at 25° C). It is insoluble in water.
Helpful?
-
-
Can Product 17850, Ciprofloxacin, be used to remove Mycoplasma contaminations?
1 answer-
We have not tested the product for that application. However, there is a reference in Roche's Biochemica Newsletter Issue 1 from 1996 that describes using Ciprofloxacin in mycoplasma contamination.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What is the potency of Product 17850, Ciprofloxacin?
1 answer-
The potency of this product has not been determined by our Quality Control or by our supplier.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service