Skip to Content
Merck
All Photos(1)

Documents

A3737

Sigma-Aldrich

Cefepime hydrochloride

Synonym(s):

Cefepime dihydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H25N6O5S2Cl · HCl · H2O
CAS Number:
Molecular Weight:
571.50
UNSPSC Code:
51282610
NACRES:
NA.85

form

powder

Quality Level

color

white to faint yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

InChI

1S/C19H24N6O5S2.2ClH.H2O/c1-25(5-3-4-6-25)7-10-8-31-17-13(16(27)24(17)14(10)18(28)29)22-15(26)12(23-30-2)11-9-32-19(20)21-11;;;/h9,13,17H,3-8H2,1-2H3,(H3-,20,21,22,26,28,29);2*1H;1H2/b23-12-;;;/t13-,17-;;;/m1.../s1

InChI key

LRAJHPGSGBRUJN-OMIVUECESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cefepime is a semi-synthetic fourth-generation cephalosporin. It is resistant to hydrolysis by common plasmid and/or chromosomally-mediated β-lactamases.

Application

Cefepime was used in antimicrobial susceptibility testing for Acinetobacter baumannii and extended-spectrum-β-lactamase-producing Escherichia coli isolated from various samples.

Biochem/physiol Actions

Cefepime is a broad-spectrum cephalosporin with activity against Gram-positive and Gram-negative bacteria. It binds to penicillin-binding proteins and disrupts the cell wall synthesis.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel J Rhodes et al.
Antimicrobial agents and chemotherapy, 58(7), 3757-3761 (2014-04-23)
While a lack of concordance is known between gold standard MIC determinations and Vitek 2, the magnitude of the discrepancy and its impact on treatment decisions for extended-spectrum-β-lactamase (ESBL)-producing Escherichia coli are not. Clinical isolates of ESBL-producing E. coli were
Kuei-Hsiang Hung et al.
Journal of clinical microbiology, 50(3), 721-726 (2011-12-23)
Heteroresistance to antimicrobial agents may affect susceptibility test results and therapeutic success. In this study, we investigated heteroresistance to cephalosporins and penicillins in Acinetobacter baumannii, a major pathogen causing nosocomial infections. Two A. baumannii isolates exhibited heteroresistance to ampicillin-sulbactam, ticarcillin-clavulanic
L B Barradell et al.
Drugs, 47(3), 471-505 (1994-03-01)
Cefepime is a 'fourth' generation cephalosporin that has a broader spectrum of antibacterial activity than the third generation cephalosporins and is more active in vitro against Gram-positive aerobic bacteria. The fact that cefepime is stable to hydrolysis by many of
M P Okamoto et al.
American journal of hospital pharmacy, 51(4), 463-477 (1994-02-15)
The chemistry, pharmacology, antimicrobial spectrum, pharmacokinetics, clinical efficacy, adverse effects, and dosage of cefepime are reviewed. Fourth-generation cephalosporins, such as cefepime, have a quaternary nitrogen that is positively charged at the 3-position, providing the properties of a zwitterion. A 2-aminothiazolyl-acetamido
Fu Peng et al.
Chinese medical journal, 127(23), 4001-4005 (2014-11-29)
Our pervious antibacterial studies on several traditional Chinese medicines have found that Patchouli oil from Pogostemon cablin had significant antibacterial activity against methicillin-resistant Staphylococcus aureus (MRSA), which has spread worldwide and infected innumerable people. In order to find the more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service