Skip to Content
Merck
All Photos(1)

Key Documents

O9639

Sigma-Aldrich

Oridonin

≥98% (HPLC), solid

Synonym(s):

7a,20-Epoxy-1a,6b,7,14-tetrahydroxy-Kaur-16-en-15-one, Isodonol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O6
CAS Number:
Molecular Weight:
364.43
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

O1[C@]2([C@]43[C@H]([C@]5([C@H]([C@@H]2O)C(CC[C@@H]5O)(C)C)C1)CC[C@H]([C@H]4O)C(=C)C3=O)O

InChI

1S/C20H28O6/c1-9-10-4-5-11-18-8-26-20(25,19(11,14(9)22)15(10)23)16(24)13(18)17(2,3)7-6-12(18)21/h10-13,15-16,21,23-25H,1,4-8H2,2-3H3/t10-,11-,12-,13+,15+,16-,18+,19-,20+/m0/s1

InChI key

SDHTXBWLVGWJFT-XKCURVIJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oridonin has potent anti-tumor activity. Oridonin targets AE (AML1-ETO) oncoprotein. Exposure to oridonin induces apoptosis in AE-bearing leukemic cells through the activation of intrinsic apoptotic pathway and triggering a caspase-3-mediated degradation of AE at D188. The compound also prolonged the lifespan of C57 mice bearing truncated AE-expressing leukemic cells without side effects like suppression of bone marrow or reduction of body weight of animals, and exerted synergic effects while combined with cytosine arabinoside. Additionally, oridonin inhibited tumor growth in nude mice inoculated with t(8;21)-harboring Kasumi-1 cells.
Oridonin has potent anti-tumor activity; targets AE oncoprotein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiang Li et al.
International journal of biological sciences, 8(6), 901-912 (2012-06-30)
Oridonin (ORI) could inhibit the proliferation and induce apoptosis in various cancer cell lines. However, the mechanism is not fully understood. Human prostate cancer (HPC) cells were cultured in vitro and cell viability was detected by the CCK-8 assay. The
Shengpeng Wang et al.
The American journal of Chinese medicine, 41(1), 177-196 (2013-01-23)
Oridonin, a natural tetracycline diterpenoid isolated from Chinese herb Rabdosia rubescens, has been reported to be a potent cytotoxic agent against a wide variety of tumors. However, its effect on highly metastatic breast cancer cells has not been addressed. In
Eric S J Harris et al.
PloS one, 7(11), e50753-e50753 (2012-12-05)
Oridonin is a diterpenoid with anti-cancer activity that occurs in the Chinese medicinal plant Isodon rubescens and some related species. While the bioactivity of oridonin has been well studied, the extent of natural variation in the production of this compound
Dandan Zheng et al.
International journal of pharmaceutics, 436(1-2), 379-386 (2012-06-27)
In this study, oridonin-loaded nanoparticles coated with galactosylated chitosan (ORI-GC-NP) were prepared for tumor targeting and their characteristics were evaluated for the morphologies, particle size and zeta potential. Oridonin-loaded nanoparticles (ORI-NP) without galactosylated chitosan were prepared as a control. The
Yang Yu et al.
Free radical research, 46(11), 1393-1405 (2012-08-14)
Oridonin, a diterpenoid compound, extracted and purified from Rabdosia rubescen has been reported to have cytotoxic effect on tumour cells through apoptosis, and tyrosine kinase pathways are involved in these processes. A specific epidermal growth factor receptor (EGFR) inhibitor AG1478

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service