Skip to Content
Merck
All Photos(2)

Key Documents

D6883

Sigma-Aldrich

2′,7′-Dichlorodihydrofluorescein diacetate

≥97% (HPLC), powder, cell-permeable non-fluorescent probe

Synonym(s):

2′,7′-Dichlorofluorescin diacetate, DCFH-DA, 2′,7′-Dichlorodihydrofluorescein diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16Cl2O7
CAS Number:
Molecular Weight:
487.29
Beilstein:
4610273
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%

Quality Level

Assay

≥97%

form

powder

storage temp.

−20°C

SMILES string

CC(=O)Oc1cc2Oc3cc(OC(C)=O)c(Cl)cc3C(c2cc1Cl)c4ccccc4C(O)=O

InChI

1S/C24H16Cl2O7/c1-11(27)31-21-9-19-15(7-17(21)25)23(13-5-3-4-6-14(13)24(29)30)16-8-18(26)22(32-12(2)28)10-20(16)33-19/h3-10,23H,1-2H3,(H,29,30)

InChI key

PXEZTIWVRVSYOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,7′-Dichlorodihydrofluorescein diacetate (DCFH-DA) is an oxidation-sensitive fluorescent dye used to detect the intracellular reactive oxygen species (ROS) level. DCFH-DA is a cell-permeable non-fluorescent probe, that can be hydrolyzed by intracellular esterase to DCFH, which forms fluorescent DCF when oxidized by the intracellular ROS.
The excitation wavelength is 504 nm and emission wavelength is 529 nm in ethanol. The use of DMSO as the solvent has not been determined, however various publications suggest that it is suitable.

Application

2′,7′-Dichlorofluorescin diacetate is suitable for use in:
  • sensitive and rapid quantitation of oxygen-reactive species in response to oxidative metabolism
  • microplate assay for detecting oxidative products in phagocytic cells
  • quantitative multiwell myeloid differentiation assay

Applications include sensitive and rapid quantitation of oxygen-reactive species in response to oxidative metabolism; microplate assay for detecting oxidative products in phagocytic cells, and quantitative multiwell myeloid differentiation assay.

Storage and Stability

This product should be stored in powdered format at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katia Cortese et al.
Life sciences, 232, 116610-116610 (2019-06-30)
The aim of this study was the characterization of the in vitro cytotoxic properties of a recently isolated diterpene compound, 7β-acetoxy-20-hydroxy-19,20-epoxyroyleanone (compound 1), extracted from Salvia corrugata, versus human cell lines. We used as model study immortalized breast epithelial cells
Pornthip Chaichompoo et al.
Journal of clinical pathology, 72(8), 520-524 (2019-04-24)
Iron overload is a major factor contributing to the overall pathology of thalassaemia, which is primarily mediated by ineffective erythropoiesis and shorter mature red blood cell (RBC) survival. Iron accumulation in RBCs generates reactive oxygen species (ROS) that cause cellular
Yuan Ji et al.
Journal of agricultural and food chemistry, 67(32), 9060-9069 (2019-07-25)
Glutathione S-transferases (GSTs) play an active role in the development of drug resistance by numerous cancer cells, including melanoma cells, which is a major cause of chemotherapy failure. As part of our continuous effort to explore why dietary polyphenols bearing
Yeonhwa Song et al.
Journal of experimental & clinical cancer research : CR, 36(1), 38-38 (2017-03-04)
CD133-positive cells in hepatocellular carcinoma (HCC) exhibit cancer stem cell (CSC)-like properties as well as resistance to chemotherapeutic agents and ionizing radiation; however, their function remains unknown. In this paper, we identified a hitherto unknown mechanism to overcome CD133-induced resistance
Shengyan Wang et al.
Electrophoresis, 40(16-17), 2204-2210 (2019-04-07)
Nanoagents achieving photodynamic therapy (PDT) and photothermal therapy (PTT) combination treatment with improved therapeutic effect are highly desirable. However, the incorporation of both PDT and PTT into a single nanoagent often requires multistep fabrication process. Herein, we report that photoactive

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service